Skip to Content
Merck
All Photos(1)

Key Documents

A5930

Sigma-Aldrich

N-Acetyl-Asp-Glu

≥97% (TLC)

Synonym(s):

N-Acetylaspartylglutamicacid, NAAG, Spaglumic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O8
CAS Number:
Molecular Weight:
304.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (TLC)

form

powder or crystals

optical activity

[α]22/D −34.5°, c = 1.1 in H2O(lit.)

storage condition

(Tightly closed. Dry)

color

white to off-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

CC(=O)NC(CC(O)=O)C(=O)NC(CCC(O)=O)C(O)=O

InChI

1S/C11H16N2O8/c1-5(14)12-7(4-9(17)18)10(19)13-6(11(20)21)2-3-8(15)16/h6-7H,2-4H2,1H3,(H,12,14)(H,13,19)(H,15,16)(H,17,18)(H,20,21)

InChI key

OPVPGKGADVGKTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

NAc-Asp-Glu

General description

Research area: Neuroscience

Application

N-Acetyl-Asp-Glu has been used in N-acetylated α-linked acidic dipeptidase (NAALADase) assay and affinity experiments.. It has also been used as a metabolite standard in liquid chromatography-mass spectrometry (LC-MS) targeted metabolomics analysis and mass-spectroscopy-based stable isotope-resolved metabolomics (SIRM) with 13C515N2-labeled-glutamine.

Biochem/physiol Actions

Endogenous neurotransmitter localized to neurons with high affinity for metabotropic glutamate receptors, mGluR3. It is an antagonist at NMDA receptors. Catabolized by carboxypeptidase II, which is expressed on astrocyte membranes, to N-acetylaspartate and glutamate.
Endogenous neurotransmitter localized to neurons with high affinity for metabotropic glutamate receptors, mGluR3; NMDA receptor antagonist.
N-Acetyl-Asp-Glu (NAAG) is an abundant neuropeptide and neurotransmitter found in the mammalian brain. NAAG acts as an agonist for metabotropic glutamate receptors, mGluR3. In addition, it also acts as an antagonist for the N-methyl d-aspartate (NMDA) receptors. It yields N-acetyl aspartate and glutamate through glutamate carboxypeptidase II (GCP II) dependent pathway. Thus, it serves as a key glutamate reservoir in cancer cells. NAAG levels in plasma could be used as a non-invasive biomarker for tumor progression.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Store desiccated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyeong Hun Lee et al.
Magnetic resonance in medicine, 82(1), 33-48 (2019-03-13)
To develop a robust method for brain metabolite quantification in proton magnetic resonance spectroscopy (1 H-MRS) using a convolutional neural network (CNN) that maps in vivo brain spectra that are typically degraded by low SNR, line broadening, and spectral baseline
Rosane Gomez et al.
Biological psychiatry, 71(3), 239-246 (2011-08-23)
Ethanol modulates glutamate and γ-aminobutyric (GABA) function. However, little is known about the acute pharmacologic effects of ethanol on levels of GABA, glutamate, and other metabolites measurable in the human cortex in vivo with proton magnetic resonance spectroscopy ((1)H-MRS). Eleven
Richard Bergeron et al.
Current medicinal chemistry, 19(9), 1360-1364 (2012-02-07)
At central synapses, glutamate is the main excitatory neurotransmitter. Once released from presynaptic terminals, glutamate activates a number of different glutamatergic receptors one of which is the ligand gated ionophore glutamatergic subtype N-methyl-D-aspartate receptors (NMDARs). NMDARs play a crucial role
Preparation and affinity identification of glutamic acid-urea small molecule analogs in prostate cancer
Zhang Z, et al.
Oncology Letters, 12(2), 1001-1006 (2016)
Yusuke Fujieda et al.
PloS one, 7(8), e43152-e43152 (2012-08-23)
Traumatic spinal cord injury (SCI) results in direct physical damage and the generation of local factors contributing to secondary pathogenesis. Untargeted metabolomic profiling was used to uncover metabolic changes and to identify relationships between metabolites and neurobehavioral functions in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service