61941
Ethylene glycol 5 M solution
Synonym(s):
Ethylene glycol solution, Additive Screening Solution 01/Kit-No 78374
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
form
liquid
Quality Level
density
1.04 g/mL at 20 °C
storage temp.
2-8°C
SMILES string
OCCO
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Ethylene glycol is a low toxicity molecule and is used for embryo cryopreservation in many domestic animals.
Ethylene glycol 5M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
Ethylene glycol 5M solution is an additive screening solution of Additive Screening Kit. Additive Screen kit is designed to allow rapid and convenient evaluation of additives and their ability to influence the crystallization of the sample. The Additive Kit provides a tool for refining crystallization conditions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2 Oral
Target Organs
Kidney
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Comparison of ethylene glycol, 1,2-propanediol and glycerol for cryopreservation of slow-cooled mouse zygotes, 4-cell embryos and blastocysts.
Emiliani S, et al.
Human Reproduction, 15, 905-905 (2000)
Andrea Marchionni et al.
ChemSusChem, 6(3), 518-528 (2013-02-14)
The electrooxidation of ethylene glycol (EG) and glycerol (G) has been studied: in alkaline media, in passive as well as active direct ethylene glycol fuel cells (DEGFCs), and in direct glycerol fuel cells (DGFCs) containing Pd-(Ni-Zn)/C as an anode electrocatalyst
Mona Sheikhi et al.
Fertility and sterility, 100(1), 170-177 (2013-04-10)
To study the preservation of follicles within ovarian tissue vitrified using only one or a combination of three permeating cryoprotectants. Experimental study. University hospital. Ovarian tissue was donated by consenting women undergoing elective cesarean section. Ovarian tissue was vitrified in
Yi Wang et al.
Journal of the American Chemical Society, 135(5), 1941-1951 (2013-01-16)
This article describes a robust method for the facile synthesis of small Ag nanocubes with edge lengths controlled in the range of 18-32 nm. The success of this new method relies on the substitution of ethylene glycol (EG)--the solvent most
Revital Goldshmid et al.
Methods (San Diego, Calif.), 84, 35-43 (2015-05-02)
A method for the preparation of suspension culture microcapsules used in the bioprocessing of human mesenchymal stem cells (hMSCs) is reported. The microcapsules are prepared from a semi-synthetic hydrogel comprising Pluronic®F127 conjugated to denatured fibrinogen. The Pluronic-fibrinogen adducts display a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service