Skip to Content
Merck
All Photos(1)

Key Documents

387614

Sigma-Aldrich

Ethylene oxide

≥99.5%

Synonym(s):

Oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
Beilstein:
102378
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

Quality Level

Assay

≥99.5%

refractive index

n20/D 1.3597 (lit.)

bp

10.7 °C (lit.)

mp

−111 °C (lit.)

density

0.882 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

C1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2

InChI key

IAYPIBMASNFSPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethylene oxide is generally synthesized by chlorohydrine process and direct oxidation of ethylene with air, catalyzed by silver(Ag).

Ethylene oxide serves as a starting material to produce ethylene glycols, surfactants, emulsifiers, and polyurethane foams.

Application

Ethylene oxide can be used as a precursor to synthesize:
  • Poly(ethylene oxides) and poly(ethyleneglycol)s.
  • Ethylene carbonate via coupling reaction with CO2 catalyzed by imidazolium zinc tetrahalides.;
  • α,ω-heterodifunctionalized poly(ethylene oxide)s using organocatalyst N-heterocyclic carbenes via ring-opening polymerization.

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Chem. Unst. Gas A - Eye Dam. 1 - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas - Repr. 1B - Skin Corr. 1B - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

N-heterocyclic carbene-organocatalyzed ring-opening polymerization of ethylene oxide in the presence of alcohols or trimethylsilyl nucleophiles as chain moderators for the synthesis of α, ω-heterodifunctionalized poly (ethylene oxide)s
Raynaud J, et al.
Macromolecules, 43(6), 2814-2823 (2010)
Multifunctional Poly (ethylene glycol)s
Obermeier B, et al.
Angewandte Chemie (International ed. in English), 50(35), 7988-7997 (2011)
Polymerization of ethylene oxide, propylene oxide, and other alkylene oxides: synthesis, novel polymer architectures, and bioconjugation
Herzberger, Jana and Niederer, et al.
Chemical Reviews, 116, 2170-2243 (2016)
Surface active ethylene oxide adducts (2013)
Non-fouling surfaces produced by gas phase pulsed plasma polymerization of an ultra low molecular weight ethylene oxide containing monomer
Wu YJ, et al.
Colloids and Surfaces. B, Biointerfaces, 18(3-4), 235-248 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service