Skip to Content
Merck
All Photos(1)

Key Documents

03747

Sigma-Aldrich

Ethylene glycol

BioUltra, ≥99.5% (GC)

Synonym(s):

1,2-Ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OH
CAS Number:
Molecular Weight:
62.07
Beilstein:
505945
EC Number:
MDL number:
UNSPSC Code:
12352137
eCl@ss:
39020501
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

vapor density

2.1 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

752 °F

expl. lim.

15.3 %

technique(s)

affinity chromatography: suitable

impurities

insoluble matter, passes filter test
≤0.002% free acid (as CH3COOH)
≤0.1% water

refractive index

n20/D 1.431 (lit.)
n20/D 1.431

bp

195-198 °C

mp

−13 °C (lit.)

density

1.113 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Al: ≤0.5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

OCCO
OCCO

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.01

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

For desorption in immuno-affinity chromatography[1][2]; Additive for the stabilization of urinary enzymes[3]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J P Li et al.
Journal of biochemical and biophysical methods, 22(4), 311-320 (1991-05-01)
An affinity chromatography column packed with nonporous agarose beads derivatized with human growth hormone via carbonyldiimidazol was used for the purification of antibodies against human growth hormone from antiserum. Desorption with 1 M sodium chloride in 60% ethylene glycol at
K Jung et al.
Clinical biochemistry, 21(1), 53-57 (1988-01-01)
We examined the stability of N-acetyl-beta-D-glucosaminidase (EC 3.2.1.30), alanine aminopeptidase (EC 3.4.11.2), alkaline phosphatase (EC 3.1.3.1), gamma-glutamyltransferase (EC 2.3.2.2), and lysozyme (EC 3.2.1.17) in urine prepared by gel filtration and supplemented with albumin, or ethylene glycol, or ethylene glycol plus
P L Lim
Molecular immunology, 24(1), 11-15 (1987-01-01)
Comparative studies on the efficacies of various buffers in eluting absorbent-bound antibodies revealed the denaturative effect of 3 M KSCN on all three IgM antibodies but not on an IgG protein, and the generally weak eluting power of glycine-HCl buffer
Peter Mygind Leth et al.
Forensic science international, 155(2-3), 179-184 (2005-10-18)
Ethylene glycol (EG) can be found in many agents, such as antifreeze. Ingestion of EG may cause serious poisoning. Adults are typically exposed when EG is ingested as a cheap substitute for ethanol or in suicide-attempts. Children may be exposed
Yi Wang et al.
Journal of the American Chemical Society, 135(5), 1941-1951 (2013-01-16)
This article describes a robust method for the facile synthesis of small Ag nanocubes with edge lengths controlled in the range of 18-32 nm. The success of this new method relies on the substitution of ethylene glycol (EG)--the solvent most

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service