Merck
  • A method for preparation of hydrogel microcapsules for stem cell bioprocessing and stem cell therapy.
A method for preparation of hydrogel microcapsules for stem cell bioprocessing and stem cell therapy.
Methods (San Diego, Calif.) (2015-05-02)
Revital Goldshmid, Iris Mironi-Harpaz, Yonatan Shachaf, Dror Seliktar
ABSTRACT
A method for the preparation of suspension culture microcapsules used in the bioprocessing of human mesenchymal stem cells (hMSCs) is reported. The microcapsules are prepared from a semi-synthetic hydrogel comprising Pluronic®F127 conjugated to denatured fibrinogen. The Pluronic-fibrinogen adducts display a lower critical solubility temperature (LCST) at ∼30 °C, thus enabling mild, cell-compatible physical crosslinking of the microcapsules in a warm gelation bath. Cell-laden microgels were prepared from a solution of Pluronic-fibrinogen hydrogel precursor and hMSCs; these were cultivated for up to 15 days in laboratory-scale suspension bioreactors and harvested by reducing the temperature of the microcapsules to disassemble the physical polymer network. The viability, proliferation and cell recovery yields of the hMSCs were shown to be better than photo-chemically crosslinked microcapsules made from a similar material. The cell culture yields, which exceeded 300% after 15 days in suspension culture, were comparable to other microcarrier systems used for the mass production of hMSCs. The simplicity of this methodology, both in terms of the cell inoculation and mild recovery conditions, represent distinct advantages for stem cell bioprocessing with suspension culture bioreactors.
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol solution, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ethylene glycol 5 M solution