Skip to Content
Merck
All Photos(4)

Key Documents

S5506

Sigma-Aldrich

Sodium nitrate

ReagentPlus®, ≥99.0%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
powder or crystals

grade

reagent

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

5.5-8 (20 °C, 50 g/L)

mp

306 °C (dec.) (lit.)

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium nitrate (chile salpeter) is a hygroscopic compound and soluble in water. It is primarily used as an oxidizing agent in organic synthesis. Additionally, sodium nitrate is used in the manufacture of glass and charcoal briquettes, and the synthesis of potassium compounds.

Application

Sodium nitrate is used:
  • To modify hydroxyapatite to prepare heterogeneous catalyst (sodium nitrate/hydroxyapatite) for Claisen–Schmidt condensation process to synthesis of chalcones.
  • In the direct acetoxylation of arenes under an anhydrous environment of TFA (trifluoroacetic acid), acetic acid, and acetic anhydride.
  • As a redox mediator in the selective oxidation of aromatic alcohols in biphase electrolysis at ambient temperature.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Fariz Ali et al.
Pathogens (Basel, Switzerland), 9(9) (2020-09-10)
To gain insight into how pathogens adapt to new hosts, Cryptococcus neoformans (H99W) was serially passaged in Galleria mellonella. The phenotypic characteristics of the passaged strain (P15) and H99W were evaluated. P15 grew faster in hemolymph than H99W, in vitro
Abhinav Sharma et al.
ACS omega, 5(8), 3924-3931 (2020-03-10)
Acute myocardial infarction (AMI), also recognized as a "heart attack," is one leading cause of death globally, and cardiac myoglobin (cMb), an important cardiac biomarker, is used for the early assessment of AMI. This paper presents an ultrasensitive, label-free electrochemical
Emilio M Ungerfeld et al.
Microorganisms, 7(5) (2019-05-01)
There is an interest in controlling rumen methanogenesis as an opportunity to both decrease the emissions of greenhouse gases and improve the energy efficiency of rumen fermentation. However, the effects of inhibiting rumen methanogenesis on fermentation are incompletely understood even
Suna Choi et al.
Journal of controlled release : official journal of the Controlled Release Society, 131(1), 70-76 (2008-08-12)
One of the most important requirements for non-viral gene delivery systems is the ability to mediate high levels of gene expression with low toxicity. After the DNA/vector complexes are taken up by cells through endocytosis, DNA is typically contained within
Joel T Kidgell et al.
International journal of biological macromolecules, 150, 839-848 (2020-02-15)
Ulvan, a sulfated polysaccharide extracted from the green seaweed genus Ulva, has bioactive properties including an immunomodulating capacity. The immunomodulatory capacity of ulvan from Ulva ohnoi, however, has not been assessed in detail. We depolymerised purified ulvan from U. ohnoi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service