Skip to Content
Merck
All Photos(3)

Documents

221341

Sigma-Aldrich

Sodium nitrate

ACS reagent, ≥99.0%

Synonym(s):

Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Agency

suitable for EPA 1621
suitable for EPA 300

Assay

≥99.0%

form

powder or crystals

impurities

≤0.005% insolubles

pH

5.5-8.3 (25 °C, 5%)

mp

306 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.001%
iodate (IO3-): ≤5 ppm
nitrite (NO2-): ≤0.001%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤3 ppm
Mg: ≥0.002%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrate is a water soluble inorganic salt that is commonly used in fertilizers. Due to its thermal stability at high temperature, it shows potential as a phase change material (PCM) for thermal energy storage.

Application

Sodium nitrate may be used for the modification of natural phosphate for use as a basic catalyst for Claisen-Schmidt condensation. It may also be used in the nitration of benzene in trifluoroacetic acid.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trifluoroacetic acid as a medium for aromatic nitration using sodium nitrate.
Spitzer UA & Stewart R.
The Journal of Organic Chemistry, 39(26), 3936-3937 (1974)
Characterization of sodium nitrate as phase change material.
Bauer T, et al.
International Journal of Thermophysics, 33(1), 91-104 (2012)
Chen Zhang et al.
Nutrients, 11(5) (2019-05-30)
The polysaccharide isolated from alfalfa was considered to be a kind of macromolecule with some biological activities; however, its molecular structure and effects on immune cells are still unclear. The objectives of this study were to explore the extraction and
Application of natural phosphate modified with sodium nitrate in the synthesis of chalcones: a soft and clean method.
Sebti S, et al.
J. Catal., 213(1), 1-6 (2003)
Polyol synthesis of platinum nanoparticles: control of morphology with sodium nitrate.
Herricks T, et al.
Nano Letters, 4(12), 2367-2371 (2004)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service