Skip to Content
Merck
All Photos(4)

Key Documents

P8394

Sigma-Aldrich

Potassium nitrate

ReagentPlus®, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
form:
powder or crystals
Pricing and availability is not currently available.

Quality Level

product line

ReagentPlus®

Assay

≥99.0%

form

powder or crystals

pH

7

mp

334 °C (lit.)

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium nitrate (KNO3) is a water-soluble inorganic salt that is commonly used as a source of nitrate.

Application

Potassium nitrate can be used in the following:      
  • KNO3/Al2O3 is used as a solid base catalyst to synthesize by vapor-phase methylation of catechol with dimethyl carbonate.
  • KNO3/cetyltrimethylammonium bromide (CTAB) catalytic system is used in the oxidation of benzyl halides to aldehydes and ketones in aqueous media.
  • In the Cu/CeO2 catalytic system, KNO3 can also be utilized as a promoter to enhance the soot oxidation activity and resistance to sulfur poisoning of the catalyst.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abraham Lin et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(6), 1802062-1802062 (2019-04-03)
Breakthroughs in cancer immunotherapies have demonstrated considerable success, though not without limitations. Non-thermal plasma (NTP) for cancer therapy has been emerging as a potential adjuvant treatment via induction of immunogenic cell death (ICD). Cancer cells undergoing ICD stimulate a patient's
Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
Jing Huang et al.
Applied and environmental microbiology, 80(1), 29-42 (2013-10-01)
Bacteria play important roles in mineral weathering and soil formation. However, few reports of mineral weathering bacteria inhabiting subsurfaces of soil profiles have been published, raising the question of whether the subsurface weathering bacteria are fundamentally distinct from those in
M A Olutoye et al.
Bioresource technology, 102(23), 10777-10783 (2011-10-11)
Fatty acid methyl esters (FAME) were produced from palm oil using eggshell modified with magnesium and potassium nitrates to form a composite, low-cost heterogeneous catalyst for transesterification. The catalyst, prepared by the combination of impregnation/co-precipitation was calcined at 830 °C

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service