Skip to Content
Merck
All Photos(1)

Key Documents

15736

Supelco

Sodium nitrate

analytical standard

Synonym(s):

E251, Chile salpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaNO3
CAS Number:
Molecular Weight:
84.99
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.995% trace metals basis

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

306 °C (dec.) (lit.)

solubility

water: soluble 874 g/L at 20 °C

cation traces

B: ≤1 mg/kg

suitability

conforms to structure for Infrared spectrum

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[O-][N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium nitrate is a nitrate salt of sodium, which finds application as a preservative in cured meat products for the protection against Clostridium botulinum and other Clostridium species.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sodium nitrate may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Ham using reversed-phase high-performance liquid chromatography with diode array detection (RP-HPLC-DAD).
  • Human saliva samples using ion chromatography (IC) technique.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce D. Craig, David S. Anderson
Handbook of Corrosion Data, 795-795 (1994)
Quantification of residual nitrite and nitrate in ham by reverse-phase high performance liquid chromatography/diode array detector
Ferreira OVLPMI and Silva S
Talanta, 74(5), 1598-1602 (2008)
Mun Fei Yam et al.
Molecules (Basel, Switzerland), 15(6), 4452-4466 (2010-07-27)
The aim of the present study was to verify the anti-inflammatory activity of Orthosiphon stamineus leaf extracts and to identify the active compound(s) contributing to its anti-inflammatory activity using a developed HPLC method. Active chloroform extract of O. stamineus was
Ion chromatographic method for simultaneous determination of nitrate and nitrite in human saliva
Helaleh HIM and Korenaga T
Journal of Chromatography. B, Biomedical Sciences and Applications, 744(2), 433-437 (2000)
Filip J Larsen et al.
The American journal of clinical nutrition, 99(4), 843-850 (2014-02-07)
Nitrate, which is an inorganic anion abundant in vegetables, increases the efficiency of isolated human mitochondria. Such an effect might be reflected in changes in the resting metabolic rate (RMR) and formation of reactive oxygen species. The bioactivation of nitrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service