Skip to Content
Merck
All Photos(3)

Key Documents

317195

Sigma-Aldrich

p-Xylene

suitable for HPLC, ≥99%

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901563
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
138 °C (lit.)
vapor pressure:
9 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.7 (vs air)

Quality Level

vapor pressure

9 mmHg ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

984 °F

expl. lim.

7 %

technique(s)

HPLC: suitable

impurities

<0.030% water

evapn. residue

<0.0005%

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

solubility

water: soluble 0.2 g/L

density

0.861 g/mL at 20 °C (lit.)

λ

H2O reference

UV absorption

λ: 294 nm Amax: 1.00
λ: 320 nm Amax: 0.10
λ: 350 nm Amax: 0.05
λ: 380-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Measurements of relative distribution constants of vaporized hydrocarbons between air and polydimethylsiloxane via capillary columns for the calculation of headspace hydrocarbon compositions.: This research demonstrates the use of p-Xylene in gas chromatography to measure the distribution constants of hydrocarbons, crucial for accurate environmental and chemical analysis (Kim & Yu, 2020).

  • Mutagenic atmospheres resulting from the photooxidation of aromatic hydrocarbon and NO(x) mixtures.: This article explores the mutagenic potential of atmospheric p-Xylene under photooxidation conditions, providing insights into air quality and health-related impacts (Riedel et al., 2018).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ultra-selective cycloaddition of dimethylfuran for renewable p-xylene with H-BEA.
Chang CC, et al.
Green Chemistry, 16(2), 585-588 (2014)
Cycloaddition of biomass-derived furans for catalytic production of renewable p-xylene.
Williams CL, et al.
ACS Catalysis, 2(6), 935-939 (2012)
Friedel-Crafts acylation of toluene and p-xylene with carboxylic acids catalyzed by zeolites.
Chiche B, et al.
The Journal of Organic Chemistry, 51(11), 2128-2130 (1986)
Metal-organic framework MIL-101 for high-resolution gas-chromatographic separation of xylene isomers and ethylbenzene.
Zhi-Yuan Gu et al.
Angewandte Chemie (International ed. in English), 49(8), 1477-1480 (2010-01-22)
G Gallastegui et al.
Bioresource technology, 102(17), 7657-7665 (2011-07-01)
Interactions of toluene and p-xylene in air treatment biofilters packed with an inert filter media were studied. The effect of the inlet load of toluene, p-xylene and mixtures of both compounds on the biodegradation rate was analyzed in three lab-scale

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service