Skip to Content
Merck
All Photos(2)

Key Documents

270709

Sigma-Aldrich

Benzene

suitable for HPLC, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein:
969212
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
80 °C (lit.)
vapor pressure:
166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

grade

HPLC grade

Quality Level

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

1043 °F

expl. lim.

8 %

technique(s)

HPLC: suitable

impurities

≤0.02% water (Karl Fischer)

evapn. residue

<0.0005%

color

APHA: ≤20

refractive index

n20/D 1.501 (lit.)

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 280 nm Amax: 1.0
λ: 290 nm Amax: 0.15
λ: 300 nm Amax: 0.06
λ: 330 nm Amax: 0.02
λ: 350-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Environmental carcinogen; hematoxin that is linked to increased incidence of leukemia in humans.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luoping Zhang et al.
Critical reviews in toxicology, 32(1), 1-42 (2002-02-16)
Benzene is an established cause of human leukemia that is thought to act by producing chromosomal aberrations and altered in cell differentiation. In several recent studies increased levels of chromosomal aberrations in peripheral blood lymphocytes were correlated with a heightened
Filamentous growth of carbon through benzene decomposition.
Oberlin A, et al.
Journal of Crystal Growth, 32(3), 335-349 (1976)
M Kiguchi et al.
Physical review letters, 101(4), 046801-046801 (2008-09-04)
Highly conductive molecular junctions were formed by direct binding of benzene molecules between two Pt electrodes. Measurements of conductance, isotopic shift in inelastic spectroscopy, and shot noise compared with calculations provide indications for a stable molecular junction where the benzene
Use of the CNDO method in spectroscopy. I. Benzene, pyridine, and the diazines.
Del Bene J and Jaffe HH.
J. Chem. Phys. , 48(44), 1807-1813 (1968)
Benzene forms hydrogen bonds with water.
Suzuki S, et al.
Science, 257(5072), 942-945 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service