Skip to Content
Merck
All Photos(2)

Key Documents

134449

Sigma-Aldrich

p-Xylene

ReagentPlus®, 99%

Synonym(s):

1,4-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901563
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/MS: suitable
bp:
138 °C (lit.)
vapor pressure:
9 mmHg ( 20 °C)

vapor density

3.7 (vs air)

Quality Level

vapor pressure

9 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

984 °F

expl. lim.

7 %

technique(s)

GC/MS: suitable

refractive index

n20/D 1.495 (lit.)

bp

138 °C (lit.)

mp

12-13 °C (lit.)

solubility

water: soluble 0.2 g/L

density

0.861 g/mL at 20 °C (lit.)

SMILES string

Cc1ccc(C)cc1

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3

InChI key

URLKBWYHVLBVBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Xylene is an isomeric form of xylene. It undergoes Friedel-Crafts acylation reaction with various acids in the presence of zeolites (catalyst). It can be obtained from the mixtures of isomeric forms of xylene and ethylbenzene isolated from the naptha fraction of petroleum. Also, it can be prepared from toluene by employing ZSM (Zeolite Socony Mobil)-5 catalysts. It is a raw material for the manufacture of PET (poly(ethylene terephthalate)) bottles. Its sustainable synthesis from ethylene and dimethylfuran in the presence of zeolite catalysts has been reported.

Application

p-Xylene may be used in the synthesis of poly-p-xylene and terephthalic acid.
p-xylene mayumb be used in the single drop microextraction of the corresponding organochlorine pesticides in vegetables, separated via gas chromatography(GC)coupled with mass spectrometry(MS).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mixed liquids for single-drop microextraction of organochlorine pesticides in vegetables
Zhang M, et al.
Talanta, 74(4), 599-604 (2008)
Friedel-Crafts acylation of toluene and p-xylene with carboxylic acids catalyzed by zeolites.
Chiche B, et al.
The Journal of Organic Chemistry, 51(11), 2128-2130 (1986)
Cycloaddition of biomass-derived furans for catalytic production of renewable p-xylene.
Williams CL, et al.
ACS Catalysis, 2(6), 935-939 (2012)
Metal-organic framework MIL-101 for high-resolution gas-chromatographic separation of xylene isomers and ethylbenzene.
Zhi-Yuan Gu et al.
Angewandte Chemie (International ed. in English), 49(8), 1477-1480 (2010-01-22)
Ultra-selective cycloaddition of dimethylfuran for renewable p-xylene with H-BEA.
Chang CC, et al.
Green Chemistry, 16(2), 585-588 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service