Skip to Content
Merck
All Photos(4)

Key Documents

220299

Sigma-Aldrich

Iron(II) chloride tetrahydrate

ReagentPlus®, 98%

Synonym(s):

Ferrous chloride tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl2 · 4H2O
CAS Number:
Molecular Weight:
198.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

10 mmHg ( 693 °C)

Quality Level

product line

ReagentPlus®

Assay

98%

form

powder, crystals or chunks

pH

2.5 (20 °C, 100 g/L)

SMILES string

O.O.O.O.Cl[Fe]Cl

InChI

1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

WSSMOXHYUFMBLS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

A variety of organically templated iron phosphates, of interest because of their rich crystal chemistry and their ability to absorb large molecules in their micropores,were prepared using this compound.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromium removal by combining the magnetic properties of iron oxide with adsorption properties of carbon nanotubes.
Gupta VK, et al.
Water Research, 45(6), 2207-2212 (2011)
The crystal structure of iron (II) chloride tetrahydrate.
Penfold BR and Grigor JA.
Acta Crystallographica, 12(11), 850854-850854 (1959)
Preparation and characterization of magnetic chitosan nanospheres.
Hritcu D, et al.
Turkish Journal of Chemistry, 33, 785-796 (2009)
Lii, K-H. et al.
Chemistry of Materials, 10, 2599-2599 (1998)
Michael Riedelberger et al.
Cell host & microbe, 27(3), 454-466 (2020-02-23)
Type I interferons (IFNs-I) fulfil multiple protective functions during pathogenic infections, but they can also cause detrimental effects and enhance immunopathology. Here, we report that IFNs-I promote the dysregulation of iron homeostasis in macrophages during systemic infections with the intracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service