Skip to Content
Merck
All Photos(2)

Key Documents

451649

Sigma-Aldrich

Iron(III) chloride

greener alternative

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Ferric chloride, Iron trichloride, Molysite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3
CAS Number:
Molecular Weight:
162.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor density

5.61 (vs air)

vapor pressure

1 mmHg ( 194 °C)

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤100.0 ppm Trace Metal Analysis

mp

304 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe/h3*1H;/q;;;+3/p-3

InChI key

RBTARNINKXHZNM-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Iron(III) chloride is brownish to black crystalline solid that is highly soluble in water. As a moderately strong Lewis acid, it highly reactive with bases and capable of forming complex ions. It is widely used as a iron precursor in the field of catalysis, solar cells and batteries.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Iron(III) chloride can be used:
  • As a precursor to fabricate nano-sized Fe-N-C catalysts for proton exchange membrane(PEM) fuel cells. FeCl3 iron precursor, helps to achieve the lowest charge transfer resistance and high peak power density.
  • As an oxidant to prepare FeOCl@PPy cathode materials for Li-ion batteries with good cycling stability.
  • As a precursor to fabricate amorphous multifunctional am-Fe–Bi/NF electrode for rechargeable Zinc-air batteries.
  • To synthesize Fe3O4@nanocellulose/TiCl nano fillers for dye-sensitized solar cells to reduce the crystallinity of the polymer.

The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resitance properties.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing graphene/CNT based electrochemical detection using magneto-nanobioprobes.
Jiang M, et al.
Material Matters, 2, 877-883 (2012)
Maria Serra et al.
Membranes, 10(5) (2020-05-15)
Corn fiber, a by-product of the starch industry, is presently incorporated in animal feed. However, it has arabinoxylans as added-value components (besides ferulic acid) that should be valorized. In this work, the raw material, a fraction enriched in arabinoxylans from
Photoredox chemistry of iron (III) chloride and iron (III) perchlorate in aqueous media. A comparative study.
David F and David PG.
The Journal of Physical Chemistry, 80(6), 579-583 (1976)
Eagleson M.
Concise Encyclopedia Chemistry, 553-553 (1994)
Priyanka Sharma et al.
Scientific reports, 2, 877-877 (2012-11-21)
Graphene and related materials have come to the forefront of research in electrochemical sensors during recent years due to the promising properties of these nanomaterials. Further applications of these nanomaterials have been hampered by insufficient sensitivity offered by these nanohybrids

Articles

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service