Skip to Content
Merck
All Photos(5)

Key Documents

215422

Sigma-Aldrich

Iron(II) sulfate heptahydrate

greener alternative
Iron(II) sulfate heptahydrate
1 of 1 reviewers received a sample product or took part in a promotion

ACS reagent, ≥99.0%

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals
Pricing and availability is not currently available.

grade

ACS reagent

vapor pressure

14.6 mmHg ( 25 °C)

Assay

≥99.0%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% insolubles
≤0.05% Substances not pptd by NH4OH

pH

3-4 (25 °C, 50 g/L)

density

1.898 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%

cation traces

Cu: ≤0.005%
Fe3+: ≤0.1%
Mn: ≤0.05%
Zn: ≤0.005%

greener alternative category

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalysis. Click here for more information.

Application

Iron(II) sulfate heptahydrate is used as a catalyst for greener amide bond formation from amines and aldehydes. It has been used in the synthesis of maghemite nanoparticles.[1]

Iron-Catalyzed Efficient Synthesis of Amides from Aldehydes and Amine Hydrochloride Salts

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of γ-Fe2O3 nanoparticles for applications in magnetic hyperthermia.
Avram A, et al.
Synthesis, 10(P151), 1-1 (2011)
Thermal analysis of iron (II) sulphate heptahydrate in air. V: Thermal decomposition of hydroxy and oxysulphates.
Swamy M and Prasad T.
Journal of Thermal Analysis and Calorimetry, 20(1), 107-114 (1981)
Thermal analysis of iron (II) sulphate heptahydrate in air: III. Thermal decomposition of intermediate hydrates.
Swami MSR and Prasad TP.
Journal of Thermal Analysis and Calorimetry, 19(2), 297-304 (1980)
Anastasia Terzopoulou et al.
Advanced healthcare materials, 9(20), e2001031-e2001031 (2020-09-10)
Microrobots and metal-organic frameworks (MOFs) have been identified as promising carriers for drug delivery applications. While clinical applications of microrobots are limited by their low drug loading efficiencies and the poor degradability of the materials used for their fabrication, MOFs
Roshini Ebenezer et al.
Tropical medicine & international health : TM & IH, 18(8), 942-951 (2013-07-11)
To assess the impact of deworming and iron supplementation on the cognitive abilities and educational achievement of school-age children in Sri Lanka. Prospective, placebo-controlled randomised study. The treatment group received deworming and weekly iron supplementation for 6 months; the control group

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I would like to know if this reagent is for cell culture

    1 answer
    1. This product has not been tested for use in cell culture. The end user would have to determine suitability. Please see the link below to review the Cell Culture Tested product options:
      https://www.sigmaaldrich.com/search/iron(ii)-sulfate-heptahydrate-cell-culture?focus=products&page=1&perpage=30&sort=relevance&term=Iron%28II%29%20sulfate%20heptahydrate%20cell%20culture&type=product

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service