Skip to Content
Merck
All Photos(5)

Key Documents

236489

Sigma-Aldrich

Iron(III) chloride hexahydrate

greener alternative

ACS reagent, 97%

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

suitable for SM 5210

vapor pressure

1 mmHg ( 194 °C)

Assay

97%
97.0-102.0% (ACS specification)

form

chunks

reaction suitability

reagent type: catalyst
core: iron

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.01% P compounds (as PO4)
≤0.01% insolubles
≤0.1% Substances not pptd. by NH4OH (as sulfates)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

anion traces

nitrate (NO3-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Cu: ≤0.003%
Fe2+:, passes test (lim. ~0.002%)
Zn: ≤0.003%

greener alternative category

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. The iron catalysts are known to be inexpensive and environmentally benign. Click here for more information.

Application

Iron(III)chloride hexahydrate can be used as a catalyst for the:
  • Esterification of fatty acids and alcohols to synthesize fatty acid esters.
  • Regioselective arylations of α-amido sulfones with electron-rich arenes to synthesize unsymmetrical or bis-symmetrical triarylmethanes via Friedel-Crafts alkylation.
  • Synthesis of acyl azide derivatives by the acylation of sodium azide with N-acylbenzotriazoles.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of γ-Fe2O3 nanoparticles for applications in magnetic hyperthermia.
Avram A, et al.
Synthesis, 10(P151), 1-1 (2011)
Crystal structure of ferric chloride hexahydrate.
Lind MD.
J. Chem. Phys. , 47(3), 990-993 (1967)
Sheng He et al.
Biomaterials, 258, 120285-120285 (2020-08-12)
Myocardial fibrosis, resulting from ischemic injury, increases tissue resistivity in the infarct area, which impedes heart synchronous electrical propagation. The uneven conduction between myocardium and fibrotic tissue leads to dys-synchronous contraction, which progresses towards ventricular dysfunction. We synthesized a conductive
Magdalena Delgado et al.
Biochemical pharmacology, 162, 213-223 (2018-12-24)
Microtubule targeting agents (MTAs) have been reported to manifest their cytotoxic effects not only in mitosis but also in interphase. However, the relationship between phase-specific susceptibility and MTA concentration, especially with respect to microtubule integrity, remains poorly defined. In addition
Lii, K-H. et al.
Chemistry of Materials, 10, 2599-2599 (1998)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service