Skip to Content
Merck
All Photos(1)

Key Documents

Y0001484

Medroxyprogesterone acetate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Medroxyprogesterone 17-acetate, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Beilstein:
2066112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

medroxyprogesterone

manufacturer/tradename

EDQM

mp

206-207 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(C)=O)C(C)=O

InChI

1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1

InChI key

PSGAAPLEWMOORI-PEINSRQWSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Medroxyprogesterone acetate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Mitsushita et al.
Gynecologic oncology, 79(1), 129-132 (2000-09-28)
Successful pregnancies after conservative progestin treatment to young women with endometrial carcinoma have recently been reported. However, it is not known for certain whether the lesion is completely eradicated in such patients. We present a case of residual endometrial carcinoma
Edith R Guilbert et al.
Contraception, 79(3), 167-177 (2009-02-03)
In the fall of 2007, the controversy about the contraceptive use of depot-medroxyprogesterone acetate (DMPA) and its potential impact on skeletal health reached the media in the province of Quebec, Canada, thereby becoming a matter of concern for the lay
Andrew M Kaunitz et al.
Contraception, 77(2), 67-76 (2008-01-30)
While depot medroxyprogesterone acetate (DMPA) is a highly effective contraceptive used by millions of women, its use is associated with bone mineral density (BMD) loss, raising concerns about long-term risk of osteoporosis and/or fractures. We conducted a systematic review of
R Kent Hermsmeyer et al.
Nature clinical practice. Cardiovascular medicine, 5(7), 387-395 (2008-06-04)
Heart disease presentation can differ between the sexes because nonobstructive coronary disease and angina unrelated to exercise are considerably more prevalent in women than in men. When the outcomes of large, randomized, controlled trials failed to demonstrate cardiac risk protection
Andrea N Simpson et al.
Gynecologic oncology, 133(2), 229-233 (2014-02-25)
Oral progestin is an alternative to hysterectomy for women with complex atypical hyperplasia (CAH) or grade one endometrial cancer (G1EC) who wish fertility preservation. We evaluated treatment efficacy and fertility outcomes in this population. Women <45 y treated with oral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service