Skip to Content
Merck
All Photos(1)

Key Documents

Y0000598

Medroxyprogesterone acetate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Medroxyprogesterone 17-acetate, 17α-Acetoxy-6α-methylprogesterone, 17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione 17-acetate, 6α-Methyl-17α-acetoxyprogesterone, 6α-Methyl-17α-hydroxyprogesterone acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34O4
CAS Number:
Molecular Weight:
386.52
Beilstein:
2066112
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

medroxyprogesterone

manufacturer/tradename

EDQM

mp

206-207 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(C)=O)C(C)=O

InChI

1S/C24H34O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h13-14,18-20H,6-12H2,1-5H3/t14-,18+,19-,20-,22+,23-,24-/m0/s1

InChI key

PSGAAPLEWMOORI-PEINSRQWSA-N

Gene Information

human ... PGR(5241)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Medroxyprogesterone acetate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert A Wild et al.
Menopause (New York, N.Y.), 20(3), 254-260 (2013-02-26)
Our objective was to determine whether metabolic syndrome (MetS) or its components modified the effect of hormone therapy (HT) on the risk of coronary heart disease (CHD) events in the Women's Health Initiative clinical trials. We performed a nested case-control
Léanie Kleynhans et al.
Infection and immunity, 81(4), 1234-1244 (2013-02-06)
The contraceptive depot medroxyprogesterone acetate (DMPA), with progestin as the single active compound, possesses selective glucocorticoid activity and can alter the expression of glucocorticoid receptor-regulated genes. We therefore propose that pharmacological doses of DMPA used for endocrine therapy could have
Richard P H Huijbregts et al.
Endocrinology, 154(3), 1282-1295 (2013-01-29)
Recent observational studies indicate an association between the use of hormonal contraceptives and acquisition and transmission of HIV-1. The biological and immunological mechanisms underlying the observed association are unknown. Depot medroxyprogesterone acetate (DMPA) is a progestin-only injectable contraceptive that is
Lee L Lanza et al.
Obstetrics and gynecology, 121(3), 593-600 (2013-05-03)
Depot medroxyprogesterone acetate (DMPA) reversibly reduces bone mineral density. To estimate the extent to which DMPA might increase fracture risk, we undertook a retrospective cohort study of fractures in DMPA users and users of non-DMPA contraceptives, using the General Practice
Jeong-Yeol Park et al.
Obstetrics and gynecology, 121(1), 136-142 (2012-12-25)
To analyze pregnancy outcomes in young women with stage IA, grade 1 endometrioid adenocarcinoma of the uterus after successful fertility-sparing management using progestin. We reviewed the medical records of 141 women with stage IA, grade 1 endometrioid adenocarcinoma of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service