Skip to Content
Merck
All Photos(1)

Key Documents

32402

Supelco

Sulfaguanidine

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(aminoiminomethyl)benzenesulfonamide, 4-Amino-N-guanylbenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N4O2S
CAS Number:
Molecular Weight:
214.24
Beilstein:
2695326
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

NC(=N)NS(=O)(=O)c1ccc(N)cc1

InChI

1S/C7H10N4O2S/c8-5-1-3-6(4-2-5)14(12,13)11-7(9)10/h1-4H,8H2,(H4,9,10,11)

InChI key

BRBKOPJOKNSWSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide
Sulfaguanidine belongs to the sulfonamide class of antibiotics. It can exhibit broad-spectrum inhibition of both gram-positive and gram-negative bacteria.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfaguanidine may be used as an analytical reference standard for the determination of sulfaguanidine in:
  • Food samples of animal origin by salting-out supported liquid extraction (SOSLE) and quick, easy, cheap, effective, rugged and safe (QuEChERS) extraction followed by analysis using nanoflow liquid chromatography high resolution mass spectrometry (LC-HRMS).
  • Bovine muscle samples by hydrophilic interaction liquid chromatography-electrospray-tandem mass spectrometry (HILIC-ESI-MS/MS) equipped with selected reaction monitoring (SRM) mode of detection.
  • Tissues, milk and eggs of food producing animals by ultrasound-assisted extraction (UAE), solid phase extraction (SPE) and LC-MS/MS.
  • Beeswax by solid-liquid extraction (SLE), liquid-liquid extraction (LLE), SPE, QuEChERS extraction and ESI-LC-MS/MS with SRM detection.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sulfonamides in beeswax by liquid chromatography coupled to tandem mass spectrometry.
Mitrowska K and Antczak M
Journal of Chromatography. B, Biomedical Sciences and Applications, 1006, 179-186 (2015)
Determination of sulfonamides in serum by on-line solid-phase extraction coupled to liquid chromatography with photoinduced fluorescence detection.
Arroyo-Manzanares N, et al.
Talanta, 138, 258-262 (2015)
Vimal K Balakrishnan et al.
Journal of chromatography. A, 1131(1-2), 1-10 (2006-08-02)
In recent years, pharmaceutical and personal care products (PPCPs) have been detected in diverse environments (including groundwater, river water, and municipal wastewater). In order to evaluate their environmental impact, PPCPs must first be accurately determined. In this study, we focused
Alan D Gift et al.
Journal of pharmaceutical sciences, 98(12), 4670-4683 (2009-05-21)
Active pharmaceutical ingredients (API) can undergo an anhydrate to hydrate transformation during wet granulation and this transformation may either result in mixed crystalline forms or an unwanted form in the final drug product. Previous studies have shown that it may
Qualitative screening of veterinary anti-microbial agents in tissues, milk, and eggs of food-producing animals using liquid chromatography coupled with tandem mass spectrometry.
Chen D, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1017, 82-88 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service