Skip to Content
Merck
All Photos(3)

Key Documents

M6013

Sigma-Aldrich

Medroxyprogesterone

Synonym(s):

17α-Hydroxy-6α-methyl-4-pregnene-3,20-dione, 17α-Hydroxy-6α-methylprogesterone, 6α-Methyl-5-pregnen-17α-ol-3,20-dione, 6-Dihydromegestrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O3
CAS Number:
Molecular Weight:
344.49
Beilstein:
2510965
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder
Assay:
≥98% (HPLC)

biological source

synthetic (organic)

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

chloroform: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](C)C3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(C)=O

InChI

1S/C22H32O3/c1-13-11-16-17(20(3)8-5-15(24)12-19(13)20)6-9-21(4)18(16)7-10-22(21,25)14(2)23/h12-13,16-18,25H,5-11H2,1-4H3/t13-,16+,17-,18-,20+,21-,22-/m0/s1

InChI key

FRQMUZJSZHZSGN-HBNHAYAOSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Caution

Much of the pharmacology literature refers to medroxyprogesterone acetate as medroxyprogesterone, but they are of course not the same substance.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adarsh J Sai et al.
Menopause (New York, N.Y.), 18(10), 1101-1112 (2011-06-30)
The aim of this study was to examine the effect of conjugated equine estrogens alone (ET), conjugated equine estrogens + medroxyprogesterone (EPT), calcitriol alone, calcitriol + EPT/ET, or placebo on serum lipid profile and analyze the interaction with estrogen receptor-α
M F Fung et al.
Obstetrics and gynecology, 78(6), 1033-1038 (1991-12-01)
We conducted a retrospective review of 44 patients with metastatic or recurrent endometrial carcinoma treated with cisplatin, doxorubicin, cyclophosphamide, and medroxyprogesterone acetate. Thirty-six women had metastatic disease; eight had recurrent disease. In the metastatic group, 12 women had positive peritoneal
Clinical significance of differences in bioavailability of medroxyprogesterone acetate preparations.
A D Stockdale et al.
Clinical pharmacokinetics, 16(3), 129-133 (1989-03-01)
The role of medroxyprogesterone acetate (MPA) in the treatment of renal adenocarcinoma.
M Kjaer
Cancer treatment reviews, 15(3), 195-209 (1988-09-01)
James Hassell et al.
Hormones and behavior, 60(2), 219-225 (2011-06-04)
Progestins and antiprogestins are widely used therapeutic agents in humans. In many cases, these are indicated for the treatment of reproductive activities. However, progesterone has widespread physiological effects including a reduction of the response to stress. We have reported that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service