Skip to Content
Merck
All Photos(2)

Key Documents

79285

Supelco

Oil Orange SS

analytical standard

Synonym(s):

Orange OT, 1-(o-Tolylazo)-2-naphthol, Solvent Orange 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4N=NC10H6OH
CAS Number:
Molecular Weight:
262.31
Beilstein:
8330630
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

124-126 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccccc1N=Nc2c(O)ccc3ccccc23

InChI

1S/C17H14N2O/c1-12-6-2-5-9-15(12)18-19-17-14-8-4-3-7-13(14)10-11-16(17)20/h2-11,20H,1H3

InChI key

BQFCCCIRTOLPEF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fujita et al.
Journal of pharmacobio-dynamics, 5(4), 259-265 (1982-04-01)
Treatment of rats with 1-m-tolueneazo-2-naphthol (mTAN) caused marked increase in liver weight, microsomal 7-ethoxycoumarin O-deethylase activity and cytochrome P-450 content, with 2 nm hypochromic shift in CO difference spectrum. It decreased NADPH-cytochrome c reductase and aminopyrine N-demethylase activities. Treatment of
O Schimmer et al.
Mutation research, 249(1), 105-110 (1991-07-01)
Seven naturally occurring furoquinoline alkaloids were investigated for their photobiological activity using arg-1 cells of Chlamydomonas reinhardtii. UV-A-mediated toxicity of the compounds was calculated from the colony-forming ability of the treated cells. The UV-A-mediated mutagenicity was measured by counting the
S Adachi et al.
Journal of pharmacobio-dynamics, 5(4), 273-277 (1982-04-01)
Effect of potent cytochrome P-448 inducer, 1-m-tolueneazo-2-naphthol (m-TAN) on hepatic microsomal UDP-glucuronyl-transferase (UDPGT) activity was studied. The UDPGT activity reached the maximum level on the fifth day after either a single injection or consecutive daily administrations. The UDPGT activities towards
Bhakti R Petigara et al.
Journal of AOAC International, 90(5), 1373-1378 (2007-10-25)
A reversed-phase liquid chromatographic method was developed to determine parts-per-million and higher levels of Sudan 1, 1-(phenylazo)-2-naphthalenol, in the disulfo monoazo color additive FD&C Yellow No. 6 and in a related monosulfo monoazo color additive, D&C Orange No. 4. Sudan
C O Mills et al.
Biochimica et biophysica acta, 876(3), 677-683 (1986-05-21)
We investigated the effect of conjugation with the aromatic amino acid tyrosine on the critical micellar concentration (CMC) of bile salts. The CMC values were determined by surface tension and by dye solubilization. The surface tension measurement employed the Du

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service