Skip to Content
Merck
All Photos(2)

Key Documents

67195

Supelco

Pigment Red 53

analytical standard

Synonym(s):

5-Chloro-2-[(2-hydroxy-1-naphthalenyl)azo]-4-methylbenzenesulfonic acid monosodium salt, Lake Red C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12ClN2NaO4S
CAS Number:
Molecular Weight:
398.80
Colour Index Number:
15585
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC1=CC=C2C(C=CC=C2)=C1/N=N/C3=CC(C)=C(Cl)C=C3S(=O)([O-])=O.[Na+]

InChI

1S/C17H13ClN2O4S.Na/c1-10-8-14(16(9-13(10)18)25(22,23)24)19-20-17-12-5-3-2-4-11(12)6-7-15(17)21;/h2-9,21H,1H3,(H,22,23,24);/q;+1/p-1/b20-19+;

InChI key

LLELVHKMCSBMCX-RZLHGTIFSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D G Goodman et al.
Journal of the National Cancer Institute, 73(1), 265-273 (1984-07-01)
In six carcinogenicity bioassays, male and female F344 rats were fed diets containing aniline hydrochloride (CAS: 142-04-1; hydrochloride benzenamide), p-chloroaniline (CAS: 106-47-8), azobenzene (CAS: 103-33-3), o-toluidine hydrochloride (CAS: 636-21-5), dapsone (CAS: 80-08-0; 4,4'-sulfonyldianiline), or D & C red No. 9
C Westmoreland et al.
Mutation research, 281(3), 163-167 (1992-03-01)
The azo-compound, D and C Red No. 9 was assayed for genotoxicity in vivo using the rat micronucleus test and the rat ex vivo liver UDS assay. Uniformly negative results were obtained in both assays, even though large oral doses
M A Weinberger et al.
Journal of the National Cancer Institute, 75(4), 681-690 (1985-10-01)
A histopathologic review of F344 rat spleens from the National Toxicology Program-National Cancer Institute bioassays of barium salt of 5-chloro-2-(2-hydroxy-1-naphthalenyl)-azo-4-methylbenzenesulfonic acid [(D & C Red No. 9) CAS: 516-00-21] and aniline HCI (CAS: 142-04-1) was conducted to assess splenotoxic changes
D&C Red No. 9 (CI Pigment Red 53:1).
IARC monographs on the evaluation of carcinogenic risks to humans, 57, 203-212 (1993-01-01)
A L Scher et al.
Journal - Association of Official Analytical Chemists, 71(5), 1007-1011 (1988-09-01)
A method is described for the determination of the intermediates in D&C Red No. 8 by reverse-phase liquid chromatography (LC). The pigment is dissolved in boiling 95% ethanol-water (1 + 1) and then precipitated. The filtrate is chromatographed by gradient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service