Skip to Content
Merck
All Photos(2)

Key Documents

80028

Supelco

Chrysoidine G

analytical standard

Synonym(s):

4-Phenylazo-m-phenylenediamine monohydrochloride, Basic Orange 2, Chrysoidine Y

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N4 · HCl
CAS Number:
Molecular Weight:
248.71
Colour Index Number:
11270
Beilstein:
3724653
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

235 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Nc1ccc(N=Nc2ccccc2)c(N)c1

InChI

1S/C12H12N4.ClH/c13-9-6-7-12(11(14)8-9)16-15-10-4-2-1-3-5-10;/h1-8H,13-14H2;1H

InChI key

MCTQNEBFZMBRSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Simultaneous determination of chrysoidine and auramine O in bean products by HPLC].
Qin Lin
Se pu = Chinese journal of chromatography, 25(5), 776-777 (2007-12-29)
Ardeshir Shokrollahi et al.
Journal of hazardous materials, 160(2-3), 435-440 (2008-04-25)
A cloud point extraction procedure was presented for the preconcentration of copper(II) ion in various samples. After complexation by 4-(phenyl diazenyl) benzene-1,3-diamine (PDBDM) (chrysoidine), copper(II) ions were quantitatively recovered in Triton X-114 after centrifugation. 0.5ml of methanol acidified with 1.0molL(-1)
G Sole et al.
Lancet (London, England), 1(8444), 1477-1479 (1985-06-29)
In a case-control study of 240 men with urothelial cancer diagnosed in 1984 and 240 matched control subjects smoking and coarse fishing were found to be significantly associated with the disease. Duration of exposure to the chrysoidine azo dyes used
M K Purkait et al.
Journal of hazardous materials, 137(2), 827-835 (2006-04-08)
Cloud point extraction (CPE) is carried out to extract chrysoidine dye from aqueous solution using two different non-ionic surfactants, TX-100 and TX-114. The effects of different operating parameters, e.g., concentrations of surfactant, dye and salt, temperature, pH on extraction of
Maggots dyed with chrysoidine.
British medical journal (Clinical research ed.), 289(6456), 1451-1452 (1984-11-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service