Skip to Content
Merck
All Photos(2)

Key Documents

861030

Sigma-Aldrich

Auramine O

Dye content 85 %, certified by the BSC

Synonym(s):

4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Basic Yellow 2, Pyoctaninum aureum

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N3 · HCl
CAS Number:
Molecular Weight:
303.83
Colour Index Number:
41000
Beilstein:
4030061
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Agency

certified by the BSC

Quality Level

form

powder or chunks

composition

Dye content, 85%

technique(s)

microbe id | staining: suitable

mp

>250 °C (dec.) (lit.)

solubility

95% ethanol: 1 mg/mL, clear to hazy, yellow to very deep yellow

λmax

370 nm
434 nm (2nd)

ε (extinction coefficient)

≥12000 at 249-255 nm in water at 0.005 g/L
≥15000 at 367-373 nm in water at 0.005 g/L
≥36000 at 429-435 nm in water at 0.005 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].CN(C)c1ccc(cc1)C(=N)c2ccc(cc2)N(C)C

InChI

1S/C17H21N3.ClH/c1-19(2)15-9-5-13(6-10-15)17(18)14-7-11-16(12-8-14)20(3)4;/h5-12,18H,1-4H3;1H

InChI key

KSCQDDRPFHTIRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Auramine O has been used to study the cuticle/cutin in the epidermis of the pollen presenter in Vangueria infausta.

Biochem/physiol Actions

Auramine O is used for the staining of acid-fast organisms, coccidia. Auramine O along with carbol generates brilliant yellow fluorochrome of tubercle bacilli. It is effective in detecting positive cases of tuberculosis. Auramine O binds to the mycolic acid in the bacterial cell wall.

Suitability

Certified for use by fluorescence microscopy in Churukian′s modification of Truant′s fluorescent method for acid fast bacilli on paraffin sections.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Garcia LS
Practical Guide to Diagnostic Parasitology (2009)
The Sigma-Aldrich Handbook of Stains, Dyes and Indicators,, 103-103 (1990)
Koneman's Color Atlas and Textbook of Diagnostic Microbiology (2006)
The epidermal cell structure of the secondary pollen presenter in Vangueria infausta (Rubiaceae: Vanguerieae) suggests a functional association with protruding onci in pollen grains.
PLoS ONE, 9, e96405-e96405 (2014)
Jessica Minion et al.
Journal of clinical microbiology, 49(5), 2024-2026 (2011-03-25)
Light-emitting diode fluorescence microscopy is being scaled up for tuberculosis control, but fading of auramine-stained slides could compromise external quality assurance. We stored auramine-stained slides and reexamined them over time. Slides stored in all environments faded quickly, with significant changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service