Skip to Content
Merck
All Photos(2)

Documents

60591

Supelco

Potassium tetrakis(4-chlorophenyl)borate

Selectophore, ≥98.0%

Synonym(s):

Tetrakis(4-chlorophenyl)boron potassium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)4BK
CAS Number:
Molecular Weight:
496.11
Beilstein:
4117827
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥98.0% (AT)
≥98.0%

form

solid

SMILES string

[K+].Clc1ccc(cc1)[B-](c2ccc(Cl)cc2)(c3ccc(Cl)cc3)c4ccc(Cl)cc4

InChI

1S/C24H16BCl4.K/c26-21-9-1-17(2-10-21)25(18-3-11-22(27)12-4-18,19-5-13-23(28)14-6-19)20-7-15-24(29)16-8-20;/h1-16H;/q-1;+1

InChI key

SAGICZRAKJSWLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Potassium tetrakis(4-chlorophenyl)borate was used as lipophilic anionic site in constructing fluorescent optode for sodium. It was also used as coating for the piezoelectric crystal in quantifying Potassium using quartz crystal microbalance.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihaela Dana Tutulea-Anastasiu et al.
Sensors (Basel, Switzerland), 13(4), 4367-4377 (2013-04-04)
All-solid-state sensors with polyvinyl chloride (PVC)-based membranes using off-the-shelf N-hydroxysuccinimide (NHS) and succinimide (Succ) ionophores were prepared using DOP (dioctyl phthalate) and NPOE (ortho-nitrophenyloctyl ether) as plasticizers. Good responses were obtained when NHS was used. The potentiometric response of the
Hacer Esra Kabak et al.
Journal of fluorescence, 29(3), 549-567 (2019-03-29)
Correct measurement of the pH in highly acidic environments is still a challenge. In such conditions most of the pH indicators suffer from instability in air or leaching from host matrices due to the solubility considerations. In this work, two
Larisa Lvova et al.
Sensors (Basel, Switzerland), 21(4) (2021-03-07)
The metalloporphyrin ligand bearing incorporated anion-exchanger fragment, 5-[4-(3-trimethylammonium)propyloxyphenyl]-10,15,20-triphenylporphyrinate of Co(II) chloride, CoTPP-N, has been tested as anion-selective ionophore in PVC-based solvent polymeric membrane sensors. A plausible sensor working mechanism includes the axial coordination of the target anion on ionophore metal
Konrad Rudnicki et al.
Analytica chimica acta, 1085, 75-84 (2019-09-17)
In this paper, the electrochemical behavior of four fluoroquinolone antibiotics (FAs) [Ciprofloxacin (Cip), Enrofloxacin (Enr), Marbofloxacin (Mar) and Ofloxacin (Ofl)] at a polarized interface between two immiscible electrolyte solutions (ITIES) has been studied using ion-transfer voltammetry (ITV). The measurements were
R C Thomas et al.
Pflugers Archiv : European journal of physiology, 390(1), 96-98 (1981-04-01)
We have developed a filling solution for silanised microelectrodes consisting of potassium tetrakis (p-chlorophenyl) borate in octanol. Microelectrodes filled with this reference liquid ion-exchanger (RLIE) have equal selectivities to Na and K, and give the same membrane potential as do

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service