Skip to Content
Merck
All Photos(2)

Key Documents

60403

Supelco

Potassium ionophore I

Selectophore, function tested

Synonym(s):

Valinomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H90N6O18
CAS Number:
Molecular Weight:
1111.32
Beilstein:
78657
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Pricing and availability is not currently available.

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

mp

184-190 °C

storage temp.

2-8°C

SMILES string

CC(C)[C@@H]1NC(=O)[C@H](C)OC(=O)[C@@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC(=O)[C@@H](NC(=O)[C@H](C)OC(=O)[C@H](NC(=O)[C@H](OC1=O)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C)C(C)C

InChI

1S/C54H90N6O18/c1-22(2)34-49(67)73-31(19)43(61)55-38(26(9)10)53(71)77-41(29(15)16)47(65)59-36(24(5)6)51(69)75-33(21)45(63)57-39(27(11)12)54(72)78-42(30(17)18)48(66)60-35(23(3)4)50(68)74-32(20)44(62)56-37(25(7)8)52(70)76-40(28(13)14)46(64)58-34/h22-42H,1-21H3,(H,55,61)(H,56,62)(H,57,63)(H,58,64)(H,59,65)(H,60,66)/t31-,32-,33-,34-,35+,36+,37-,38-,39-,40+,41+,42+/m0/s1

InChI key

FCFNRCROJUBPLU-RPUZOQEISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Important research tool for membrane studies[1]; As ionophor in membrane electrodes for the determination of K+-activities[2][3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of potassium ion concentration in serum using a highly selective liquid-membrane electrode.
L A Pioda et al.
Clinica chimica acta; international journal of clinical chemistry, 29(2), 289-293 (1970-08-01)
H F Osswald et al.
Clinical chemistry, 25(1), 39-43 (1979-01-01)
We describe a flow-through system with an ion-selective electrode for measurement of blood potassium ion concentration, continuously and on-line off the extracorporeal blood circulation in an operating theater during human open-heart surgery. Comparison measurements were made with the SMA flame
Model membranes.
A D Bangham
Chemistry and physics of lipids, 8(4), 386-392 (1972-05-01)
Viktor K Jirsa et al.
Brain : a journal of neurology, 137(Pt 8), 2210-2230 (2014-06-13)
Seizures can occur spontaneously and in a recurrent manner, which defines epilepsy; or they can be induced in a normal brain under a variety of conditions in most neuronal networks and species from flies to humans. Such universality raises the
Ko-Shing Chang et al.
Biosensors & bioelectronics, 31(1), 137-143 (2011-11-01)
A silicon nanowire field-effect transistor (SiNW-FET) coated with a polyvinyl chloride (PVC) membrane containing valinomycin (VAL) was employed as a biosensor (referred to as VAL-PVC/SiNW-FET) to detect the K(+)-efflux from live chromaffin cells. The detection sensitivity of K(+) with the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service