Skip to Content
Merck
All Photos(2)

Key Documents

87255

Supelco

Tetradodecylammonium tetrakis(4-chlorophenyl)borate

for ion-selective electrodes, Selectophore

Synonym(s):

ETH 500, Tetrakis(4-chlorophenyl)borate tetradodecylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H116BCl4N
CAS Number:
Molecular Weight:
1148.32
Beilstein:
8894894
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

mp

79-80 °C

SMILES string

Clc1ccc(cc1)[B-](c2ccc(Cl)cc2)(c3ccc(Cl)cc3)c4ccc(Cl)cc4.CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C48H100N.C24H16BCl4/c1-5-9-13-17-21-25-29-33-37-41-45-49(46-42-38-34-30-26-22-18-14-10-6-2,47-43-39-35-31-27-23-19-15-11-7-3)48-44-40-36-32-28-24-20-16-12-8-4;26-21-9-1-17(2-10-21)25(18-3-11-22(27)12-4-18,19-5-13-23(28)14-6-19)20-7-15-24(29)16-8-20/h5-48H2,1-4H3;1-16H/q+1;-1

InChI key

OKKRVCFTMNLHTL-UHFFFAOYSA-N

General description

ETH 500 is a lipophilic electrolyte mostly used as an anionic additive.
Visit our Sensor Applications portal to learn more.

Application

ETH 500 has been used in the preparation of:
  • molecularly imprinted polymers, which can be used during potentiometric sensing method for determining neutral phenols.
  • copolymer Ca2+ selective membrane cocktail during electrochemical impedance spectroscopic analysis to study the water intake and layer formation in ion-selective electrode.

Other Notes

Highly lipophilic additive for membranes of neutral carrier-based ion-selective macroelectrodes; Additive in membranes of optodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric sensor for determination of neutral bisphenol A using a molecularly imprinted polymer as a receptor.
Kou LJ
Analytical and Bioanalytical Chemistry, 405(14), 4931-4936 (2013)
Ahmed Galal Eldin et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-12)
Two novel all-solid-state potentiometric sensors for the determination of azide ion are prepared and described here for the first time. The sensors are based on the use of iron II-phthalocyanine (Fe-PC) neutral carrier complex and nitron-azide ion-pair complex (Nit-N₃-) as
D. Ammann et al.
Analytica Chimica Acta, 171, 119-119 (1985)
W.E. Morf et al.
Pure and Applied Chemistry. Chimie Pure Et Appliquee, 61, 1613-1613 (1989)
Lipophilic and immobilized anionic additives in solvent polymeric membranes of cation-selective chemical sensors.
Rosatzin, Thomas, et al.
Analytica Chimica Acta, 280.2, 197-208 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service