Skip to Content
Merck
All Photos(2)

Documents

21193

Supelco

Calcium ionophore II

Selectophore, function tested

Synonym(s):

N,N,N′,N′-Tetra[cyclohexyl]diglycolic acid diamide, N,N,N′,N′-Tetracyclohexyl-3-oxapentanediamide, ETH 129

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48N2O3
CAS Number:
Molecular Weight:
460.69
Beilstein:
6491651
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

solid

quality

function tested

SMILES string

O=C(COCC(=O)N(C1CCCCC1)C2CCCCC2)N(C3CCCCC3)C4CCCCC4

InChI

1S/C28H48N2O3/c31-27(29(23-13-5-1-6-14-23)24-15-7-2-8-16-24)21-33-22-28(32)30(25-17-9-3-10-18-25)26-19-11-4-12-20-26/h23-26H,1-22H2

InChI key

URAUKAJXWWFQSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Calcium ionophore II (ETH 129) was used in preparing calcium selective minielectrodes which are used in determination of dissociation constant (Kd).
Suitable for intracellular calcium measurements.
Lipophilic ionophore for Ca2+-selective electrodes with detection limit in the sub-nanomolar range. Extremely efficient carrier of Ca2+-ions; induces a selectivity in membranes for Ca2+ over Mg2+ by a factor of about 104.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Hove-Madsen et al.
Biophysical journal, 63(1), 89-97 (1992-07-01)
We have examined the binding of the fluorescent Ca indicator indo-1 to cellular protein in permeabilized ventricular myocytes and also to soluble and particulate myocyte protein. Using either a filtration technique or equilibrium dialysis, and conditions similar to those in
Scott T Keene et al.
Advanced healthcare materials, 8(24), e1901321-e1901321 (2019-11-13)
Wearable health monitoring has garnered considerable interest from the healthcare industry as an evolutionary alternative to standard practices with the ability to provide rapid, off-site diagnosis and patient-monitoring. In particular, sweat-based wearable biosensors offer a noninvasive route to continuously monitor
Patrick C Bradshaw et al.
BMC biochemistry, 7, 4-4 (2006-02-08)
Divalent cations are required for many essential functions of mitochondrial metabolism. Yet the transporters that mediate the flux of these molecules into and out of the mitochondrion remain largely unknown. Previous studies in yeast have led to the molecular identification
Xiaoxue Liu et al.
Mikrochimica acta, 187(9), 525-525 (2020-08-29)
Ultrathin metallic WS2 (M-WS2) nanosheets and semiconductive WS2 (S-WS2) nanosheets were exfoliated and for the first time employed as ion-to-electron transducing layer to construct an all-solid-state ion-selective electrode. Importantly, we found that the transducing efficiency of WS2 nanosheet-based solid-contact layer is
Yi Yuan et al.
Analytical chemistry, 76(18), 5570-5578 (2004-09-15)
Chronoamperometry was carried out at liquid/liquid interfaces supported at the tip of micropipet electrodes for direct determination of the diffusion coefficient of a species in the outer solution. The diffusion coefficient was used for subsequent determination of the transferred charges

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service