Skip to Content
Merck
All Photos(1)

Key Documents

23131

Sigma-Aldrich

Ethyl chloroformate

purum, ≥98.0% (GC)

Synonym(s):

Chloroformic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOC2H5
CAS Number:
Molecular Weight:
108.52
Beilstein:
385653
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.74 (vs air)

Quality Level

vapor pressure

3.42 psi ( 20 °C)

grade

purum

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

842 °F

refractive index

n20/D 1.395 (lit.)
n20/D 1.396

bp

93 °C (lit.)

mp

−81 °C (lit.)

density

1.139 g/mL at 20 °C
1.135 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

CCOC(Cl)=O

InChI

1S/C3H5ClO2/c1-2-6-3(4)5/h2H2,1H3

InChI key

RIFGWPKJUGCATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • GC-MS Analysis of Resveratrol Isomers in Red Wine: Application of ethyl chloroformate for derivatization of resveratrol isomers, enhancing GC-MS analysis effectiveness, crucial for quality control in food chemistry and drug discovery focusing on dietary supplements (Di Fabio et al., 2020).
  • Determination of Free Aromatic Carboxylic Acids and Phenols in Juices: Uses ethyl chloroformate for rapid GC-MS analysis, important for food safety and pharmacology by verifying phenolic bioactive compounds in consumer products (Incocciati et al., 2021).
  • Synthesis of Diaryl Ketones: Discusses a nickel-catalyzed method of synthesizing diaryl ketones using ethyl chloroformate, valuable for material science chemists developing new materials and pharmaceutical compounds (Shi & Hu, 2019).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man-Jeong Paik et al.
Journal of chromatography. A, 1214(1-2), 151-156 (2008-11-11)
N-ethoxycarbonylation was combined with (S)-1-phenylethylamidation for enantioseparation of amino acids by gas chromatography (GC) and gas chromatography-mass spectrometry (GC-MS) on achiral capillary columns. The method provided complete enantioseparations of 12 amino acids as diastereomeric N-ethoxycarbonyl/(S)-1-phenylethylamides with exceptional resolutions for proline
Xiumei Tao et al.
Analytical and bioanalytical chemistry, 391(8), 2881-2889 (2008-07-16)
An optimized method based on GC-MS with ethyl chloroformate derivatization has been developed for the comprehensive analysis of endogenous metabolites in serum. Twenty-two reference standards and serum samples were used to validate the proposed method. The correlation coefficient was higher
Guo-Xiang Xie et al.
Journal of pharmaceutical and biomedical analysis, 43(3), 920-925 (2006-10-13)
Dencichine (beta-N-oxalyl-l-alpha,beta-diaminopropionic acid) is a haemostatic agent present in well-known traditional Chinese medicinal herbs such as Panax notoginseng, as well as other Panax species. It is also a reported neurotoxic agent found in Lathyrus sativus (grass pea seed) and cycad
P Husek et al.
Journal of chromatography. B, Biomedical sciences and applications, 693(2), 499-502 (1997-06-06)
Following deproteinization of plasma with organic solvents the supernatant was shaken with hexane and cation-exchange resin in an Eppendorf tube to remove fatty and amino acids and the medium was subjected to direct treatment with ethyl chloroformate under catalytic influence
Gary G Chen et al.
Journal of mass spectrometry : JMS, 44(8), 1203-1210 (2009-06-11)
A quantitative method for putrescine (PUT), spermidine (SPD) and spermine (SPM) in homogenized postmortem human brain tissue is described that employs a novel, simple and rapid extractive derivatization with ethylchloroformate and trifluoroacetylation. These amines are metabolites of ornithine and are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service