Skip to Content
Merck
All Photos(2)

Key Documents

8.00881

Sigma-Aldrich

Ethyl chloroformate

for synthesis

Synonym(s):

Ethyl chloroformate, Ethyl chlorocarbonate, Ethoxycarbonyl chloride, Chloroformic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOC2H5
CAS Number:
Molecular Weight:
108.52
Beilstein:
385653
MDL number:
UNSPSC Code:
12352100
EC Index Number:
208-778-5
NACRES:
NA.22
grade:
for synthesis
synthesis grade
bp:
93 °C (lit.)
vapor pressure:
3.42 psi ( 20 °C)
54 hPa ( 20 °C)

grade

for synthesis
synthesis grade

Quality Level

vapor density

3.74 (vs air)

vapor pressure

3.42 psi ( 20 °C)
54 hPa ( 20 °C)

form

liquid

autoignition temp.

450 °C
842 °F

potency

204 mg/kg LD50, oral (Rat)
7120 mg/kg LD50, skin (Rabbit)

expl. lim.

3.7-12.6 % (v/v)

dilution

(for synthesis)

refractive index

n20/D 1.395 (lit.)

bp

93 °C (lit.)

mp

-81 °C
−81 °C (lit.)

transition temp

flash point 16 °C

density

1.14 g/cm3 at 20 °C
1.135 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

CCOC(Cl)=O

InChI

1S/C3H5ClO2/c1-2-6-3(4)5/h2H2,1H3

InChI key

RIFGWPKJUGCATF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for synthesis

Application

Ethyl chloroformate for synthesis. CAS 541-41-3, EC Number 208-778-5, chemical formula ClCOOC₂H₅.

Analysis Note

Assay (GC, area%)≥ 98.0 % (a/a)Density (d 20 °C/ 4 °C)1.138 - 1.140Identity (IR)passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service