Skip to Content
Merck
All Photos(1)

Documents

177989

Sigma-Aldrich

Isobutyl chloroformate

98%

Synonym(s):

Chloroformic acid isobutyl ester, IBCF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
136.58
Beilstein:
956590
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.33 mmHg ( 20 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.407 (lit.)

bp

128.8 °C (lit.)

solubility

benzene: miscible
chloroform: miscible
diethyl ether: miscible

density

1.053 g/mL at 25 °C (lit.)

SMILES string

CC(C)COC(Cl)=O

InChI

1S/C5H9ClO2/c1-4(2)3-8-5(6)7/h4H,3H2,1-2H3

InChI key

YOETUEMZNOLGDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobutyl chloroformate is used as a peptide reagent. In oligonucleotide synthesis, IBCF blocks the 5′-hydroxyl function of deoxyribosides. It also serves as condensing reagent.

Application

  • Preparation of a volatile derivative of taurine and application to gas chromatographic determination of urinary taurine.: Demonstrates the utility of isobutyl chloroformate in preparing volatile derivatives of taurine for gas chromatographic analysis, offering a methodological advancement in clinical biochemistry (Masuoka et al., 1989).
  • Quinazoline antifolates inhibiting thymidylate synthase: synthesis of four oligo(L-gamma-glutamyl) conjugates of N10-propargyl-5,8-dideazafolic acid and their enzyme inhibition.: This article investigates the synthesis and biological activity of quinazoline antifolates, using techniques including isobutyl chloroformate, relevant in medicinal chemistry and drug development (Pawelczak et al., 1989).
  • Coupling of peptides to protein carriers by mixed anhydride procedure.: Discusses a novel technique using isobutyl chloroformate for peptide coupling to proteins, useful in bioconjugate chemistry and vaccine development (Samokhin & Filimonov, 1985).
  • Folate analogues altered in the C9-N10 bridge region. 18. Synthesis and antitumor evaluation of 11-oxahomoaminopterin and related compounds.: Investigates the role of isobutyl chloroformate in synthesizing folate analogues for cancer research, showing its importance in therapeutic chemistry (Nair et al., 1981).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Chromatography. B, Biomedical Sciences and Applications, 617, 191-191 (1993)
Isobutyl Chloroformate
Ray T
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
A new family of highly potent inhibitors of microbes: synthesis and conjugation of elastin based peptides to piperazine derivative
Suhas R, et al.
International Journal of Peptide Research and Therapeutics, 18(2), 89-98 (2012)
Veronika Dufková et al.
Journal of chromatography. A, 1216(49), 8659-8664 (2009-10-28)
A rapid and simple derivatization procedure has been developed for gas chromatographic determination of perfluorinated organic acids (PFCAs, C(6)-C(12)), using isobutyl chloroformate (IBCF) to convert the acids into the more volatile isobutyl esters, under catalysis by pyridine. The procedure was
C H Paik et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 24(10), 932-936 (1983-10-01)
Diethylenetriaminepentaacetic acid (DTPA) was conjugated with antibody to human serum albumin (Ab) at low concentration (300 micrograms/ml, 2.0 microM) via the DTPA carboxycarbonyl mixed-anhydride method. To study parameters determining the balance between the degree of conjugation and the antibody-binding activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service