Skip to Content
Merck
All Photos(3)

Documents

190306

Sigma-Aldrich

Diisobutylaluminum hydride solution

1.0 M in hexanes

Synonym(s):

DIBAL, DIBAL-H

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]2AlH
CAS Number:
Molecular Weight:
142.22
Beilstein:
4123663
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in hexanes

density

0.701 g/mL at 25 °C

SMILES string

CC(C)C[AlH]CC(C)C

InChI

1S/2C4H9.Al.H/c2*1-4(2)3;;/h2*4H,1H2,2-3H3;;

InChI key

AZWXAPCAJCYGIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutylaluminum hydride reducing agent is commonly used for the reduction of an ester to aldehyde.

Application

Diisobutylaluminum hydride solution (1.0 M in hexanes) can be used in the synthesis of potassium diisobutyl-t-butoxyaluminum hydride (PDBBA), a novel reducing agent for the chemoselective conversion of esters in the presence of nitriles. The DIBAL-triethylamine complex can reduce α-oxoketene dithioacetal to the saturated ketone.
Used in Pd-catalyzed reductive debromination of secondary alkyl bromides. O-Debenzylation and ring opening of perbenzylated furanosides. Convenient in situ generation of HZrCp2Cl from ZrCp2Cl2 and DIBAL-H.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

alpha.-Oxoketene dithioacetal chemistry. 2. Conjugate reductions with electrophilic reducing agents.
Gammill R
The Journal of Organic Chemistry, 46(17), 3555-3558 (1981)
Chemoselective Reduction of Esters to Aldehydes by Potassium Diisobutyl-t-butoxyaluminum Hydride (PDBBA).
Chae MJ, et al.
Bull. Korean Chem. Soc., 28(12), 2517-2517 (2007)
J Marco-Contelles et al.
Carbohydrate research, 335(1), 63-70 (2001-09-13)
The reaction of DIBALH with bis(heteroannulated)-pyranosides containing the perhydrofuro[2,3-b]pyran moiety is described. The hydride attack at the anomeric carbon (C-9a) resulted in the exclusive tetrahydrofuran ring opening. The selectivity of this reaction has been evaluated as other benzylidene acetals built
D J Kopecky et al.
The Journal of organic chemistry, 65(1), 191-198 (2000-05-18)
An optimized protocol for the DIBALH reductive acetylation of acyclic esters and diesters is described. This reductive acetylation procedure allows a wide variety of esters to be converted into the corresponding alpha-acetoxy ethers in good to excellent yields. It was
Y Kitade et al.
Nucleic acids symposium series, (27)(27), 107-108 (1992-01-01)
Reaction of purine nucleosides, such as 2',3'-isopropylideneinosine (1a) and 2',3'-isopropylideneadenosine (1c), with diisobutylaluminum hydride (DIBAL) in dry tetrahydrofurane resulted in the formation of the corresponding 9-(2',3'-isopropylideneribity)purines (2) in good yields. Oxidation of the ribityl derivatives (2) with NalO4 and subsequent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service