07295
Nitrate Ionophore VI
Selectophore™, function tested
Synonym(s):
9-Hexadecyl-1,7,11,17-tetraoxa-2,6,12,16-tetraazacycloeicosane
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C28H60N4O4
CAS Number:
Molecular Weight:
516.80
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Recommended Products
grade
for ion-selective electrodes
Quality Level
product line
Selectophore™
quality
function tested
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCC1CONCCCNOCCCONCCCNOC1
InChI
1S/C28H60N4O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19-28-26-35-31-22-16-20-29-33-24-18-25-34-30-21-17-23-32-36-27-28/h28-32H,2-27H2,1H3
InChI key
NQESDIIOYWZCLK-UHFFFAOYSA-N
Related Categories
General description
New ionophore for online monitoring of nitrate obtains superior characteristics over all existing nitrate ionophores.
Visit our Sensor Applications portal to learn more.
Application
Selected application examples
Detection limit: <200μg NO3-/L
Ion selectivity: logKpot(NO3/NO2):-1.2, logKpot(NO3/Cl):-2.5, logKpot(NO3/Br):-1.1, logKpot(NO3/HCO3):-3.2, logKpot(NO3/HPO4):-4.4, logKpot(NO3/SO4):-4.5
Nitrate ionophore VI may be used in polyvinyl chloride/dioctylsebacate (PVC/DOS) matrix using a highly sensitive piezoresitive silicon oxide cantilevers for determination of nitrates in soil.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Components
Recommended Membrane Composition
5.2 wt% Nitrate ionophore VI (07295)
47.1 wt% Dibutyl phthalate (80100)
0.6 wt% Tetraoctylammonium chloride (87991)
47.1 wt% Poly(vinyl chloride) high molecular weight (81392)
5.2 wt% Nitrate ionophore VI (07295)
47.1 wt% Dibutyl phthalate (80100)
0.6 wt% Tetraoctylammonium chloride (87991)
47.1 wt% Poly(vinyl chloride) high molecular weight (81392)
Physical properties
Lipophilicity: log P = 8.3
Legal Information
Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ayokunle O Olanrewaju et al.
ACS sensors, 5(4), 952-959 (2020-04-07)
Poor adherence to pre-exposure prophylaxis (PrEP) and antiretroviral therapy (ART) can lead to human immunodeficiency virus (HIV) acquisition and emergence of drug-resistant infections, respectively. Measurement of antiviral drug levels provides objective adherence information that may help prevent adverse health outcomes.
David M Fialho et al.
Chemical reviews, 120(11), 4806-4830 (2020-05-19)
The origin of nucleotides is a major question in origins-of-life research. Given the central importance of RNA in biology and the influential RNA World hypothesis, a great deal of this research has focused on finding possible prebiotic syntheses of the
Ayokunle O Olanrewaju et al.
ACS sensors, 5(4), 952-959 (2020-04-07)
Poor adherence to pre-exposure prophylaxis (PrEP) and antiretroviral therapy (ART) can lead to human immunodeficiency virus (HIV) acquisition and emergence of drug-resistant infections, respectively. Measurement of antiviral drug levels provides objective adherence information that may help prevent adverse health outcomes.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service