Skip to Content
Merck
All Photos(1)

Documents

MAB3806-C

Sigma-Aldrich

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, Ascites Free

clone 10H11.E12, from mouse

Synonym(s):

Serine-protein kinase ATM, Ser1981 phosphorylated, Ataxia telangiectasia mutated, Ser1981 phosphorylated, A-T mutated, Ser1981 phosphorylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10H11.E12, monoclonal

species reactivity

mouse, human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer1981)

Gene Information

human ... ATM(472)

General description

Serine-protein kinase ATM (EC 2.7.11.1; UniProt Q13315; also known as A-T mutated, Ataxia telangiectasia mutated) is encoded by the ATM (also known as AT) gene (Gene ID 472) in human. Ataxia telangiectasia mutated kinase (ATM) and ataxia telangiectasia and Rad3-related kinase (ATR) are related kinases that regulate cell cycle checkpoints and DNA repair. Mutation in the ATM gene results in the autosomal recessive disease ataxia telangiectasia (AT). Known ATM substrates include p53, p95/NBS1, MDM2, Chk2, BRCA1, CtIP, 4E-BP1, and Chk1. The S/TQ sequence constitutes the essential substrates phosphorylation site motif. Hydrophobic amino acids at positions -3 and -1, and negatively charged amino acids at position +1 are positive determinants for substrate phosphorylation, while positively charged residues surrounding the S/TQ are negative determinants. The complex phenotypes of cells derived from patients with AT suggests that ATM has additional cellular substrates. ATM is present as an inactive homodimer or multimer prior to activation. DNA double-stranded breaks induced by ionizing radiation (IR) cause rapid ATM autophosphorylation at Ser1981 in human or the Ser1987 equivalent in mouse.

Specificity

Reacts with ATM Kinase phosphorylated at serine 1981. Clone 10H11.E12 is covered by US patent No. 6,916,627 and 7,108,992.

Immunogen

Epitope: pSer1981.
Synthetic peptide corresponding to a.a. 1974-1988 of human ATM with phosphorylated Ser1981 (SLAFEEG[pS]QSTTISS).

Application

Anti-phospho-ATM (Ser1981) Antibody, clone 10H11.E12, Ascites Free is an antibody against phospho-ATM for use in Immunocytochemistry, Immunoprecipitation, ELISA, Western Blotting.
Immunoprecipitation Analysis: A representative lot detected Ser1981-phosphorylated ATM interaction with endogenous EphA5 by co-immunoprecipitation using chromatin-enriched protein fractions of irradiated NCI-H460 human lung cancer cells (Staquicini, F.I., et al. (2015). J. Biol. Chem. 290(12):7345-7359).
ELISA Analysis: a representative lot was employed as the capture antibody for the detection of interaction between EphA5 and Ser1981-phosphorylated ATM. ATM pSer1981 captured from NCI-H460 human lung cancer cell extracts interacted with recombinant EphA5 cytoplasmic domain, but not with EphA5 extracellular domain or kinase domain (Staquicini, F.I., et al. (2015). J. Biol. Chem. 290(12):7345-7359).
Immunocytochemistry Analysis: A representative lot detected radiation-/IR-induced upregulation of nuclear ATM pSer1981 foci by fluorescent immunocytochemistry staining of methanol-fixed NCI-H460 human lung cancer cells (Staquicini, F.I., et al. (2015). J. Biol. Chem. 290(12):7345-7359).
Immunocytochemistry Analysis: A representative lot detected radiation-/IR-induced ATM Ser1981 phosphorylation by fluorescent immunocytochemistry staining of 1.2% formaldehyde-fixed, 0.1% Triton X-100-permeabilized HT29 cells (Bardelle, C., and Boros, J. (2012). J. Biomol. Screen. 17(7):912-920).
Immunocytochemistry Analysis: A representative lot detected ionizing radiation-/IR-induced ATM Ser1981 phosphorylation by fluorescent immunocytochemistry staining of methanol/acetone-fixed primary human foreskin fibroblasts (HFFs) (Bakkenist, C.J., et al. (2004). Cancer Res. 64(11):3748-3752).
Western Blotting Analysis: A representative lot detected epirubicin-induced ATM Ser1981 (Ser1987 in mouse) phosphorylation in mouse embryonic fibroblasts (MEFs) and human MCF-7 cells (Khongkow, P., et al. (2014). Oncogene. 33(32): 4144-4155).
Western Blotting Analysis: A representative lot detected camptothethin-induced ATM Ser1987 (Ser1981 in human) phosphorylation in primary mouse cortical neurons (Brochier, C., et al. (2013). J. Neurosci. 33(20): 8621-8632).
Western Blotting Analysis: A representative lot detected radiation-/IR-induced ATM Ser1981 phosphorylation in HT29, HeLa, and HEK293T cells (Bardelle, C., and Boros, J. (2012). J. Biomol. Screen. 17(7):912-920).
Western Blotting Analysis: A representative lot detected ionizing radiation-/IR-induced ATM Ser1981 phosphorylation in U2OS cells by Western blotting using whole cell lyates or B56γ immunoprecipitate (Shouse, G.P., et al. (2011). Oncogene. 30(35):3755-3765).
Western Blotting Analysis: A representative lot detected ionizing radiation-/IR-induced ATM Ser1981 phosphorylation in cultured primary human foreskin fibroblasts (HFFs), as well as passage-dependent basal ATM Ser1981 phosphorylation levels in HFFs (Bakkenist, C.J., et al. (2004). Cancer Res. 64(11):3748-3752).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: 4.0 µg/mL of this antibody detected Camptothecin (Cat. No. 208925) treatment-induced increase of nuclear ATM pSer1981 foci in HeLa cells.

Target description

350.7 kDa calculated.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Leung et al.
Frontiers in neuroscience, 17, 1225226-1225226 (2023-08-28)
Emerging evidence suggests cellular senescence, as a consequence of excess DNA damage and deficient repair, to be a driver of brain dysfunction following repeated mild traumatic brain injury (rmTBI). This study aimed to further investigate the role of deficient DNA
Alena Svobodová Kovaříková et al.
Epigenetics & chromatin, 16(1), 26-26 (2023-06-16)
RNA modifications have been known for many years, but their function has not been fully elucidated yet. For instance, the regulatory role of acetylation on N4-cytidine (ac4C) in RNA can be explored not only in terms of RNA stability and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service