Skip to Content
Merck
All Photos(1)

Key Documents

56741

Supelco

IGEPAL® CA-630

Synonym(s):

Nonidet P 40, Nonylphenyl-polyethylenglycol, Octylphenoxy poly(ethyleneoxy)ethanol, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)nC14H22O
CAS Number:
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25
Pricing and availability is not currently available.

description

non-ionic

Quality Level

form

liquid

mol wt

~603.0 g/mol

refractive index

n20/D 1.492

CMC

0.08 mM (20-25°C)

transition temp

cloud point 63-67  °C

density

1.06 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤1000 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤1000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

HLB

13

SMILES string

O(CCO)c1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

IGEPAL® CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes. It is chemically indistinguishable from Nonidet P-40.[1]

Application

IGEPAL® CA-630 has been used:
  • In phosducin co-precipitation assay for pull down of Epitope-Tagged HAGγ1 and HAGγ1C71S[2]
  • As a component of lysis buffer for cell culture and sample preparation for MAPK (mitogen-activated protein kinases) analysis, Immunoblotting[3][4]
  • In the extraction of cells in Oil Red O staining procedure to visualise lipid formation in cells during adipogenesis[4]
  • As a component of lysis buffer containing Tris-HCl, NaCl and EDTA for immunoblotting experiment[5]

Other Notes

Nonionic, non-denaturing detergent

Quality

Chemically indistinguishable from Nonidet P-40, which is no longer commercially available.

Caution

May become turbid on storage; a clear product can be obtained on heating to 40°C.
May develop turbidity or sediment during storage. A clear liquid can be obtained on heating to 40°C.

Legal Information

IGEPAL is a registered trademark of Solvay
Nonidet is a trademark of Shell Chemical Co.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

483.8 °F - closed cup

Flash Point(C)

251 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Regulation of ERRa gene expression by estrogen receptor agonists and antagonists in SKBR3 breast cancer cells: differential molecular mechanisms mediated by G protein-coupled receptor GPR30/GPER-1.
Li Y, et al.
Molecular Endocrinology, 24(5), 969-980 (2010)
Farnesylation of the Transducin G Protein Gamma Subunit Is a Prerequisite for Its Ciliary Targeting in Rod Photoreceptors.
Frontiers in Molecular Neuroscience, 11, 16-16 (2018)
MEK?ERK and heparin-susceptible signaling pathways are involved in cell-cycle entry of the wound edge retinal pigment epithelium cells in the adult newt.
Yoshikawa T, et al.
Pigment Cell & Melanoma Research, 25(1), 66-82 (2012)
Selective mutations in estrogen receptor alpha' s D-domain alters nuclear translocation and non-ERE gene regulatory mechanisms.
Burns K A, et al.
The Journal of Biological Chemistry, jbc-M110 (2011)
Arun K Mishra et al.
Molecular microbiology, 80(5), 1241-1259 (2011-03-26)
Lipomannan (LM) and lipoarabinomannan (LAM) are key Corynebacterineae glycoconjugates that are integral components of the mycobacterial cell wall, and are potent immunomodulators during infection. LAM is a complex heteropolysaccharide synthesized by an array of essential glycosyltransferase family C (GT-C) members

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. what is the concentration of this IGEPAL® CA-630 solution ?

    1 answer
    1. This product is considered a neat chemical, so you may assume 100% in your calculations.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service