Skip to Content
Merck
All Photos(1)

Documents

AB1284

Sigma-Aldrich

Anti-Heme Oxygenase 1 Antibody

Chemicon®, from rabbit

Synonym(s):

HO-1, HSP32

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, mouse, rat

should not react with

canine

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Hmox1(513221)
dog ... Hmox1(442987)
mouse ... Hmox1(15368)
rat ... Hmox1(24451)

Specificity

Recognizes Heme oxygenase-1.

Immunogen

A synthetic peptide with the amino acid sequence H-D-L-S-E-A-L-K-E-A-T-K-E-V-H-C-NH2 corresponding to amino acid residues [12-25] + Cys-NH2 of the human HO-1 protein (Yoshida et al., 1988).

Application

Research Category
Metabolism
Research Sub Category
Toxicology & Drug Resistance
This Anti-Heme Oxygenase 1 Antibody is validated for use in IH, WB for the detection of Heme Oxygenase 1.
Western Blot: 1:1,000 (using ECL). A single band of ~32 kD is observed. Reactivity can be completely abolished by pre-adsorption of the antibody with the complementary peptide (Catalog Number AG253). Also available are microsomal tissue preparations for control purposes (Rat spleen microsomal preparation, Catalog Number AG257).

Immunohistochemistry: 1:50-1:1,000 on human airway biopsy, human spleen, human kidney and porcine arteries.

Optimal working dilutions must be determined by the end user.

Target description

32 kDa

Physical form

Affinity Purified immunoglobulin. Liquid in PBS containing 0.01M sodium azide.
ImmunoAffinity Purified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Lung

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Cuadrado et al.
The Journal of biological chemistry, 289(22), 15244-15258 (2014-04-25)
The small GTPase protein RAC1 participates in innate immunity by activating a complex program that includes cytoskeleton remodeling, chemotaxis, activation of NADPH oxidase, and modulation of gene expression. However, its role in regulating the transcriptional signatures that in term control
Gamze Cebi et al.
Renal failure, 38(9), 1554-1559 (2016-11-04)
Myoglobinuric acute renal failure (MARF) may develop after severe muscle injury. Heme oxygenase-1 (HO-1), a stress-response protein, has been implicated as a protective agent against MARF. We hypothesized that hyperbaric oxygen therapy (HBOT) may alleviate MARF by inducing renal HO-1
Isabel Lastres-Becker et al.
Biomolecules, 12(9) (2022-09-24)
Amyotrophic lateral sclerosis (ALS) and frontotemporal dementia (FTD) are related neurodegenerative disorders displaying substantial overlay, although there are substantial differences at the molecular level. Currently, there is no effective treatment for these diseases. The transcription factor NRF2 has been postulated
Shelley X L Zhang et al.
American journal of respiratory cell and molecular biology, 40(3), 332-339 (2008-09-09)
Whole-body hypoxic preconditioning (WHPC) prolongs survival of mice exposed to severe hypoxia by attenuating pulmonary edema and preserving gas exchange. However, the cellular and molecular mechanism(s) of this protection remains unclear. The objective of this study was to identify the
Fulvio Santacatterina et al.
Oncotarget, 7(1), 490-508 (2015-11-26)
The ATPase Inhibitory Factor 1 (IF1) is an inhibitor of the mitochondrial H+-ATP synthase that regulates the activity of both oxidative phosphorylation (OXPHOS) and cell death. Here, we have developed transgenic Tet-On and Tet-Off mice that express a mutant active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service