Skip to Content
Merck
All Photos(8)

Documents

D5637

Sigma-Aldrich

3,3′-Diaminobenzidine tetrahydrochloride hydrate

≥96%

Synonym(s):

(1,1′-Biphenyl)-3,3′,4,4′-tetramine tetrahydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H14N4 · 4HCl · xH2O
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

≥96%

form

powder, crystals or chunks

technique(s)

titration: 96-104% (with NaOH)

color

white to light brown, and Faint Red and Faint Grey to Light Grey

mp

280 °C

solubility

H2O: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

O.Cl.Cl.Cl.Cl.Nc1ccc(cc1N)-c2ccc(N)c(N)c2

InChI

1S/C12H14N4.4ClH.H2O/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;;/h1-6H,13-16H2;4*1H;1H2

InChI key

DXWSCXIZIHILNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,3′-Diaminobenzidine tetrahydrochloride hydrate is a chromogen commonly used in immunohistochemistry and other staining techniques to visualize the presence of antigens or antibodies in biological samples. DAB HCl is designed as an immunohistology stain to visualize peroxidase activity in cells. DAB oxidation can be measured using a spectrophotometer.

Application

3,3′-Diaminobenzidine tetrahydrochloride hydrate is suitable for immunohistology staining procedures, western blotting, and FISH (fluorescence in situ hybridization). DAB is used in studying epithelial cell apoptosis in the event of early cataracts, and immunohistochemistry protocols of autosomal dominant polycystic kidney disease (PKD).

Biochem/physiol Actions

In the peroxidase reaction, DAB serves as a hydrogen donor in the presence of peroxide. The oxidized DAB forms an insoluble brown-colored complex.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franc Llorens et al.
Nature communications, 11(1), 619-619 (2020-02-01)
The clinical diagnosis of vascular dementia (VaD) is based on imaging criteria, and specific biochemical markers are not available. Here, we investigated the potential of cerebrospinal fluid (CSF) lipocalin 2 (LCN2), a secreted glycoprotein that has been suggested as mediating
Nicole M Derks et al.
Neuroscience research, 66(1), 117-123 (2009-10-17)
Women have higher vulnerability to stress and stress-induced diseases than men. Estrogen may be involved in the control of sex-dependent stress adaptation via estrogen receptors alpha and beta (ERalpha/beta). Urocortin 1 (Ucn1) in the npEW plays an important role in
Fatima Solange Pasini et al.
Journal of gastroenterology, 49(11), 1453-1466 (2013-11-13)
The TNM Classification of Malignant Tumours (TNM) staging system is the primary means of determining a prognosis for gastric adenocarcinoma (GC). However, tumor behavior in the individual patient is unpredictable and in spite of treatment advances, a classification of 'advanced
Jooske F van Lidth de Jeude et al.
Cancer research, 78(21), 6098-6106 (2018-09-21)
Deletion of endoplasmic reticulum resident chaperone Grp78 results in activation of the unfolded protein response and causes rapid depletion of the entire intestinal epithelium. Whether modest reduction of Grp78 may affect stem cell fate without compromising intestinal integrity remains unknown.
Daniel Balazs Fulop et al.
Scientific reports, 9(1), 14598-14598 (2019-10-12)
Pituitary adenylate cyclase activating polypeptide (PACAP) is a regulatory and cytoprotective neuropeptide, its deficiency implies accelerated aging in mice. It is present in the auditory system having antiapoptotic effects. Expression of Ca2+-binding proteins and its PAC1 receptor differs in the

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service