926965
Gold coated silica nanoparticles
1 wt. % in water, 600 nm (non-porous silica), 20 nm (gold nanoparticles)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
concentration
1 wt. % in water
particle size
20 nm (gold nanoparticles)
600 nm (non-porous silica)
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Gold coated silica nanoparticles, are composed of a spherical silica core coated with gold nanoparticles on the surface. These materials have unique chemical and optical properties make them suitable and attractive for applications in, medicine, batteries, catalysis, sensors and spectroscopy. Gold coated silica nanoparticles have material a large surface area, non-porous silica, excellent charge transport, and biocompatibility.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Biosensors & bioelectronics, 153, 112054-112054 (2020-02-15)
Hybrid silica-gold based sensors show attractive performance in sensing technologies. Due to their interesting optical properties and biological compatibility, gold nanoparticles (AuNPs) have been extensively implemented in sensing technology. Hybridization of AuNPs with silica NPs as a material with unique
Talanta, 184, 537-556 (2018-04-21)
Recent advances in nanotechnology are as a result of the development of engineered nanoparticles. Efficiently, metallic nanoparticles have been widely exploited for biomedical application and among them, gold nanoparticles (AuNPs) are highly remarkable. Consequent upon their significant nature, spherical and
A novel method for the synthesis of monodisperse gold-coated silica nanoparticles.
Journal of Nanoparticle Research, 14, 650-650 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service