Skip to Content
MilliporeSigma

Skip To

M403000

CUTAG CPG

pore size 1000 Å, 25-35 μmol/g

Synonym(s):

CUTAG CPG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$141.00
10 g
Check Cart for Availability
$1,200.00

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200

$141.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Proligo Reagents

form

solid

pore size

1000 Å

storage temp.

2-8°C

General description

CUTAG CPG from Proligo® Reagents is a universal CPG, i.e. comprises a universal linker ("UnyLinker"). It can be utilized as a substitute for traditional nucleoside-loaded CPG during the synthesis of any oligonucleotide sequence, regardless of the 3’-end nucleoside.The CUTAG CPG can be utilized under standard conditions and reagents used for synthesizing oligonucleotides with nucleoside-loaded CPG. The key distinction in using Proligo Reagent’s CUTAG CPG for oligonucleotide preparation is that the 3’-base of the oligonucleotide to be synthesized is not connected to the support. This base needs to be incorporated in the initial synthesis cycle on the CUTAG support.

Features and Benefits

CUTAG CPG can substitute for all conventional DNA and RNA supports. It is compatible with most of the oligonucleotide modifiers described in the literature and so can also be employed to produce 3’-modified- or thioated oligonucleotides.

Legal Information

This product is covered by US Patent 7,202,264 owned by Isis Pharmaceuticals, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M802000M3030005.05894
product line

Proligo Reagents

product line

-

product line

Proligo Reagents

product line

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

solid

form

-

form

-

form

solid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

pore size

1000 Å

pore size

2000 Å

pore size

500 Å

pore size

-


Still not finding the right product?

Explore all of our products under CUTAG CPG


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the meaning of 25-35 umole/gm

    1 answer
    1. The specification of 25-35 µmol/g refers to the functional group. Essentially, there will be 25 to 35 micromoles of the functional group per gram of controlled pore glass (CPG). This is somewhat indicative of the yield, however, as the length of the oligo increases the yield per gram would predictably decrease.

      See the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/427/414/proligo-cutag-cpg-user-instructions.pdf

      Helpful?

Reviews

No rating value

Active Filters