Skip to Content
Merck
All Photos(1)

Key Documents

W340804

Sigma-Aldrich

Methyl dihydrojasmonate, mixture of cis and trans

≥96%, FG

Synonym(s):

(3-Oxo-2-pentylcyclopentyl)acetic acid methyl ester, Methyl (3-oxo-2-pentylcyclopentyl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22O3
CAS Number:
Molecular Weight:
226.31
FEMA Number:
3408
EC Number:
Council of Europe no.:
10785
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.520
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA

Assay

≥96%

refractive index

n20/D 1.459 (lit.)

bp

110 °C/0.2 mmHg (lit.)

solubility

H2O: insoluble
paraffin: soluble
propylene glycol: soluble

density

0.998 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

jasmine; floral

SMILES string

CCCCCC1C(CCC1=O)CC(=O)OC

InChI

1S/C13H22O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h10-11H,3-9H2,1-2H3

InChI key

KVWWIYGFBYDJQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl dihydrojasmonate is a fragrance ingredient mainly used in the perfumes, fragrance formulations, personal care and cosmetic products. It occurs naturally in pine honey and rhododendron honey.

Other Notes

Natural occurrence: Jasmine

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile compounds of pine honeys
Silici S.
Turkish Journal of Biology, 35(5), 641-645 (2011)
Fragrance material review on methyl dihydrojasmonate.
Scognamiglio J, et al.
Food And Chemical Toxicology, 50, S562-S571 (2012)
Characterization of volatile compounds of Rhododendron honey.
Silici S.
Mellifera, 10(19) (2010)
RIFM fragrance ingredient safety assessment, methyl dihydrojasmonate, CAS registry number 24851-98-7.
Api AM, et al.
Food And Chemical Toxicology, 82, S114-S121 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service