Skip to Content
Merck
All Photos(1)

Documents

W341002

Sigma-Aldrich

Methyl jasmonate

≥98%, stabilized, FG

Synonym(s):

3-Oxo-2-(2-pentenyl)cyclopentaneacetic acid, methyl ester, Methyl 3-oxo-2-(2-pentenyl)cyclopentaneacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C5H6(CH2CH=CHC2H5)CH2CO2CH3
CAS Number:
Molecular Weight:
224.30
FEMA Number:
3410
EC Number:
Council of Europe no.:
10821
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.521
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

≥98%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.474 (lit.)

bp

110 °C/0.2 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; jasmine; floral; fruity; sweet

SMILES string

CC\C=C\CC1C(CCC1=O)CC(=O)OC

InChI

1S/C13H20O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h4-5,10-11H,3,6-9H2,1-2H3/b5-4+

InChI key

GEWDNTWNSAZUDX-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Methyl jasmonate is a plant growth regulator mainly used to enhance the aroma quality of some fruits and vegetables.

Application

Possible application (Flavor): Has many flavor uses including plum, peach, apricot and tutti-frutti.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolong Hao et al.
Biotechnology and applied biochemistry, 62(1), 24-31 (2014-05-02)
Tanshinone is a group of active diterpenes, which are widely used in the treatment of cardiovascular disease. In this study, methyl jasmonate (MJ) and salicylic acid (SA) were used to investigate their effects on tanshinone accumulation and biosynthetic gene expression
Arabidopsis mutants selected for resistance to the phytotoxin coronatine are male sterile, insensitive to methyl jasmonate, and resistant to a bacterial pathogen.
Feys BJ, et al.
Plant Cell, 6(5), 751-759 (1994)
Bjørn Dueholm et al.
BMC evolutionary biology, 15, 122-122 (2015-06-27)
Large proliferations of cytochrome P450 encoding genes resulting from gene duplications can be termed as 'blooms', providing genetic material for the genesis and evolution of biosynthetic pathways. Furanocoumarins are allelochemicals produced by many of the species in Apiaceaous plants belonging
Qiumei Liao et al.
IET nanobiotechnology, 9(1), 35-42 (2015-02-05)
The fluorescence labelling of plant hormone binding sites is an important analytical technique in research on the molecular mechanisms of plant hormone activities. The authors synthesised a jasmonic acid (JA)-conjugated ZnS:Mn quantum dot (QD) probe, with a cubic structure and
Aroma changes of black tea prepared from methyl jasmonate treated tea plants.
Shi J, et al.
Journal of Zhejiang University. Science. B, 15(4), 313-313 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service