Skip to Content
Merck
All Photos(1)

Key Documents

W310700

Sigma-Aldrich

Vanillin

≥97%, FCC, FG

Synonym(s):

4-Hydroxy-3-methoxybenzaldehyde, Vanillic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CHO
CAS Number:
Molecular Weight:
152.15
FEMA Number:
3107
Beilstein:
472792
EC Number:
Council of Europe no.:
107
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.018
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 182.60
FDA 21 CFR 182.90

vapor density

5.3 (vs air)

vapor pressure

>0.01 mmHg ( 25 °C)

description

Agency/Method; meets purity specifications of JECFA

Assay

≥97%

form

powder or crystals

bp

170 °C/15 mmHg (lit.)

mp

81-83 °C (lit.)

solubility

DMSO: soluble(lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; sweet; vanilla

SMILES string

COc1cc(C=O)ccc1O

InChI

1S/C8H8O3/c1-11-8-4-6(5-9)2-3-7(8)10/h2-5,10H,1H3

InChI key

MWOOGOJBHIARFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillin is the world′s most popular flavor and fragrance compound.[1] Vanillin, a polar aromatic flavor compound, is present in the extracts of natural and artificial vanilla flavors.[2]

Application


  • Vanillin and 4-hydroxybenzyl alcohol promotes cell proliferation and neuroblast differentiation in the dentate gyrus of mice via the increase of brain-derived neurotrophic factor and tropomyosin-related kinase B.: The study explored how vanillin and 4-hydroxybenzyl alcohol promote cell proliferation and neuroblast differentiation in mice, highlighting their potential neurogenic effects (Cho et al., 2016).

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

319.6 - 321.4 °F - closed cup

Flash Point(C)

159.8 - 160.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nethaji J Gallage et al.
Molecular plant, 8(1), 40-57 (2015-01-13)
In recent years, biotechnology-derived production of flavors and fragrances has expanded rapidly. The world's most popular flavor, vanillin, is no exception. This review outlines the current state of biotechnology-based vanillin synthesis with the use of ferulic acid, eugenol, and glucose
Kriengsak Lirdprapamongkol et al.
In vivo (Athens, Greece), 24(4), 501-506 (2010-07-30)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a promising anticancer agent which selectively kills cancer cells with little effect on normal cells. However, TRAIL resistance is widely found in cancer cells. We have previously reported antimetatstatic and antiangiogenic effects of
Subhashini Selvaraju et al.
Chemical communications (Cambridge, England), 51(14), 2957-2959 (2015-01-17)
An eumelanin-inspired core derived from the natural product, vanillin (vanilla bean extract) was utilized for the synthesis of eumelanin-inspired small molecules and polymer via Sonogashira cross coupling. The materials demonstrate that the methyl 4,7-dibromo-5,6-dimethoxy-N-methyl-1H-indole-2-carboxylate core can serve as a new
Determination of vanillin and related flavor compounds in natural vanilla extracts and vanilla-flavored foods by thin layer chromatography and automated multiple development.
Belay MT and Poole CF.
Chromatographia, 37(7-8), 365-373 (1993)
Jinyong Xu et al.
Psychiatry research, 225(3), 509-514 (2015-01-18)
Olfaction plays an important role in emotions in our daily life. Pleasant odors are known to evoke positive emotions, inducing relaxation and calmness. The beneficial effects of vanillin on depressive model rats were investigated using a combination of behavioral assessments

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service