Skip to Content
Merck
All Photos(2)

Key Documents

392707

Sigma-Aldrich

Methyl jasmonate

95%

Synonym(s):

3-Oxo-2-(2-pentenyl)cyclopentaneacetic acid, methyl ester, Methyl 3-oxo-2-(2-pentenyl)cyclopentaneacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O=)C5H6(CH2CH=CHC2H5)CH2CO2CH3
CAS Number:
Molecular Weight:
224.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

contains

Synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.474 (lit.)

bp

110 °C/0.2 mmHg (lit.)

solubility

water: soluble 340 mg/L at 25 °C(lit.)

density

1.03 g/mL at 25 °C (lit.)

functional group

ester
ketone

SMILES string

CC\C=C\CC1C(CCC1=O)CC(=O)OC

InChI

1S/C13H20O3/c1-3-4-5-6-11-10(7-8-12(11)14)9-13(15)16-2/h4-5,10-11H,3,6-9H2,1-2H3/b5-4+

InChI key

GEWDNTWNSAZUDX-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl jasmonate (MJ, MeJA), a methyl ester of jasmonic acid (JA) is a plant growth regulator. It is a fragrant volatile compound isolated from the flowers of Jasminum grandiflorum. It has been reported to play an important role in the plant′s response to pathogens and wound.

Application

Methyl jasmonate (MJ, MeJA) is suitable in controlling blue mold decay in sweet cherry fruit caused by Penicillium expansum.

Methyl jasmonate may be used in the following studies:
  • As a potential anti-cancer agent that shows selective cytotoxic effect towards cancer cells.
  • To induce the defensive proteinase inhibitor proteins synthesis in plant leaves.
  • As an elicitor to enrich the total anthocyanin content (TAC) in radish sprouts.
  • As an elicitor in inducing the biosynthesis of trans-resveratrol, a plant phenol in Vitis vinifera cv. Negramaro cell cultures.
  • As a mediator of extensive plant transcriptome reprogramming/ remodeling on exogenous treatment to Salvia sclarea leaves.
  • As a modulator in the expression of chalcone synthase (chs) and proline-rich cell wall protein (PRP), two wound-responsive genes in soybean suspension cultures.
  • As a starting material in the synthesis of [13C,2H3]-MeJA, an internal standard used in the quantitative determination of MeJA in plant tissues.
  • As a test compound in the application of reduced graphene oxide–poly(safranine T) film on glassy carbon electrode (rGO–PST/GCE) in the electrochemical determination of MeJA in jasmine essential oil.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of Reduced Graphene Oxide-Poly (Safranine T) Film via One-Step Polymerization for Electrochemical Determination of Methyl Jasmonate.
Sun J, et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 23(8), 701-708 (2015)
Methyl Jasmonate Primed Defense Responses Against Penicillium expansum in Sweet Cherry Fruit.
Wang Lei, et al.
Plant Molecular Biology Reporter, 1-8 (2015)
Annett Richter et al.
The Plant journal : for cell and molecular biology, 106(1), 245-257 (2021-01-19)
The maize (Zea mays) genome encodes three indole-3-glycerolphosphate synthase enzymes (IGPS1, 2, and 3) catalyzing the conversion of 1-(2-carboxyphenylamino)-l-deoxyribulose-5-phosphate to indole-3-glycerolphosphate. Three further maize enzymes (BX1, benzoxazinoneless 1; TSA, tryptophan synthase alpha subunit; and IGL, indole glycerolphosphate lyase) convert indole-3-glycerolphosphate
Yang Yu et al.
Plant biotechnology journal, 18(3), 679-690 (2019-08-17)
Plant defence is multilayered and is essential for surviving in a changing environment. The discovery of long noncoding RNAs (lncRNAs) has dramatically extended our understanding of post-transcriptional gene regulation in diverse biological processes. However, the expression profile and function of
E E Farmer et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(19), 7713-7716 (1990-10-01)
Inducible defensive responses in plants are known to be activated locally and systemically by signaling molecules that are produced at sites of pathogen or insect attacks, but only one chemical signal, ethylene, is known to travel through the atmosphere to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service