Skip to Content
Merck
All Photos(1)

Documents

W288403

Sigma-Aldrich

1-Phenyl-1-propanol

≥97%, FG

Synonym(s):

α-Ethylbenzyl alcohol, (±)-1-Phenylpropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(C6H5)OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
2884
Beilstein:
1906759
EC Number:
Council of Europe no.:
82
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39023126
PubChem Substance ID:
Flavis number:
2.033
NACRES:
NA.21

biological source

synthetic

grade

FG

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.52 (lit.)

bp

103 °C/14 mmHg (lit.)

density

0.994 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; floral; sweet

SMILES string

CCC(O)c1ccccc1

InChI

1S/C9H12O/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3

InChI key

DYUQAZSOFZSPHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Phenyl-1-propanol is a secondary alcohol that can be used as a flavoring agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of Food and Color Additives, 1, 2196-2196 (1997)
Stefan Ottiger et al.
Journal of chromatography. A, 1162(1), 74-82 (2007-02-17)
The separation of the enantiomers of 1-phenyl-1-propanol by supercritical fluid chromatography on a chiral stationary phase, which consists of cellulose tris (3,5-dimethylphenylcarbamate) coated on a silica support (Chiralcel-OD), is studied under overloaded, non-linear chromatographic conditions. Pulse experiments are performed at
Arvind Rajendran et al.
Journal of chromatography. A, 1092(1), 55-64 (2005-09-29)
The enantioseparation of 1-phenyl-1-propanol through the supercritical fluid-simulated moving bed (SF-SMB) process is studied. Non-linear isotherms were measured on an analytical column, and used together with the triangle theory for SMB design to select operating conditions for the SF-SMB. Experiments
Arvind Rajendran et al.
Journal of chromatography. A, 1076(1-2), 183-188 (2005-06-25)
The supercritical fluid chromatography (SFC) separation of the enantiomers of 1-phenyl-1-propanol on the chiral stationary phase Chiralcel OD under linear conditions is studied. Supercritical CO2 modified with methanol is used as a mobile phase. The effect of modifier concentration, pressure
Anna M Costa et al.
Journal of the American Chemical Society, 124(24), 6929-6941 (2002-06-13)
The optimization of asymmetric catalysts for enantioselective synthesis has conventionally revolved around the synthesis and screening of enantiopure ligands. In contrast, we have optimized an asymmetric reaction by modification of a series of achiral ligands. Thus, employing (S)-3,3'-diphenyl BINOL [(S)-Ph(2)-BINOL]

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service