Skip to Content
Merck
All Photos(1)

Documents

16350

Sigma-Aldrich

Bromobenzene

≥99.5% (GC)

Synonym(s):

1-Bromobenzene, Bromobenzol, Monobromobenzene, Phenyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Br
CAS Number:
Molecular Weight:
157.01
Beilstein:
1236661
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5.41 (vs air)

vapor pressure

10 mmHg ( 40 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1051 °F

expl. lim.

36.5 %

refractive index

n20/D 1.559 (lit.)
n20/D 1.559

bp

156 °C (lit.)

mp

−31 °C (lit.)

solubility

alcohol: soluble 10.4g/100g at 25 °C
diethyl ether: soluble 71.3g/100g at 25 °C
water: insoluble 0.045g/100g at 30 °C (practically)
benzene: miscible
chloroform: miscible
hydrocarbons: miscible (petr.)

density

1.491 g/mL at 25 °C (lit.)

SMILES string

Brc1ccccc1

InChI

1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H

InChI key

QARVLSVVCXYDNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Bromobenzene was used in the synthesis of four-armed star chain transfer agent required for preparation of amphiphilic star graft copolymers.

Biochem/physiol Actions

Bromobenzene induces hepatic necrosis via the formation of a reactive metabolite that arylates vital cellular macromolecules.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Star-like PAA-g-PPO well-defined amphiphilic graft copolymer synthesized by ATNRC and SET-NRC reaction.
Li Y, et al.
Journal of Polymer Science: Part A, General Papers, 48(10), 2084-2097 (2010)
Bromobenzene-induced liver necrosis. Protective role of glutathione and evidence for 3,4-bromobenzene oxide as the hepatotoxic metabolite.
D J Jollow et al.
Pharmacology, 11(3), 151-169 (1974-01-01)
Yuning Ma et al.
Environmental science & technology, 46(24), 13112-13117 (2012-11-28)
The 2,4,6-tribromophenoxy moiety is a common structural feature of several brominated flame retardants, and we have previously reported on the environmental concentrations of one such compound, 1,2-bis(2,4,6-tribromophenoxy) ethane (TBE). Here we report the atmospheric concentrations of TBE and three other
Verónica Guilarte et al.
Organic letters, 13(19), 5100-5103 (2011-09-03)
An efficient synthesis of thiophenes and benzo[b]thiophenes has been developed from easily available bromoenynes and o-alkynylbromobenzene derivatives. This novel one-pot procedure involves a Pd-catalyzed C-S bond formation using a hydrogen sulfide surrogate followed by a heterocyclization reaction. Moreover, in situ
Sriram Gopi et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48(8-9), 2170-2175 (2010-05-22)
The objective of the current study was to investigate the protective effect of an aqueous extract of Phyllanthus fraternus (AEPF) against bromobenzene induced mitochondrial dysfunction in rat liver mitochondria. Administration of bromobenzene (10 mmol/kg body wt.) significantly decreased the rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service