Skip to Content
Merck
All Photos(1)

Documents

179817

Sigma-Aldrich

2-Phenyl-1-propanol

97%

Synonym(s):

(±)-2-Phenyl-1-propanol, β-Methylphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)CH2OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1906760
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.526 (lit.)

bp

110-111 °C/10 mmHg (lit.)

density

0.975 g/mL at 25 °C (lit.)

SMILES string

CC(CO)c1ccccc1

InChI

1S/C9H12O/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3

InChI key

RNDNSYIPLPAXAZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The mass spectra of 2-phenyl-1-propanol was studied by positive- and negative-ion-fast atom bombardment-mass spectrometric ionization technique.

Application

2-Phenyl-1-propanol was used to study the synergism between enzyme catalysis and microwave irradiation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ganapati D Yadav et al.
Bioresource technology, 109, 1-6 (2012-02-07)
Lipase catalyzed transesterification was investigated to study the synergistic effect of microwave irradiation and enzyme catalysis. Transesterification of ethyl-3-phenylpropanoate with n-butanol was chosen as the model reaction using immobilized enzymes such as Novozyme 435, Lipozyme RMIM and Lipozyme TL IM
I F Gaunt et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 20(5), 519-525 (1982-10-01)
Phenylpropan-1-ol was added to the diet of groups of 15 male and 15 female rats to provide intakes of 0 (control), 10, 40 or 160 mg/kg/day for 13 wk. No effects on body weight, food intake, water intake, haematology, semi-quantitative
K R Henne et al.
Biochemistry, 40(29), 8597-8605 (2001-07-18)
The active site topography of rabbit CYP4B1 has been studied relative to CYP2B1 and CYP102 using a variety of aromatic probe substrates. Oxidation of the prochiral substrate cumene by CYP4B1, but not CYP2B1 or CYP102, resulted in the formation of
W Gielsdorf et al.
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 89(3), 191-195 (1982-01-01)
The FAB (Fast Atom Bombardment)-mass spectrometric ionization technique, which has now been available for about 1 year, has been successfully employed in forensic toxicology. The mass spectral behaviour of some representative drug-glucuronides (Codeine, p-Nitrophenol and 2-Phenyl-1-propanol) were studied by positive-
S Goenechea et al.
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 97(2), 83-88 (1986-01-01)
The behaviour of 2-phenyl-1-propanol (I) and 2-phenyl-2-propanol (II) and their glucuronides with HCl has been investigated. While I shows a high acidic constancy, II undergoes a partial conversion into 2-phenylpropane (III) which itself yields numerous products. The glucosidic bond of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service