Skip to Content
Merck
All Photos(1)

Key Documents

184756

Sigma-Aldrich

4-Phenyl-1-butanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)4OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
2042122
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

140-142 °C/14 mmHg (lit.)

density

0.984 g/mL at 20 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

OCCCCc1ccccc1

InChI

1S/C10H14O/c11-9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7,11H,4-5,8-9H2

InChI key

LDZLXQFDGRCELX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenyl-1-butanol is oxidized by ceric ammonium nitrate in acetonitrile to afford 2-phenyltetrahydrofuran. It undergoes cyclization in presence of phosphoric acid at high temperature to yield tetralin.

Application

4-Phenyl-1-butanol was used in synthesis of NK105, a paclitaxel-incorporating micellar nanoparticle formulation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fabrice Gritti et al.
Journal of chromatography. A, 1524, 108-120 (2017-10-11)
A twin-column recycling separation process (TCRSP) is assembled and used to generate higher speed and/or higher resolution levels than those of the usual non-recycling process at the same back pressure. It enables the users to solve very challenging separation problems
New Friedel-Crafts chemistry. XIX. Cyclialkylations of some phenylalkanols.
Khalaf AA and Roberts RM.
The Journal of Organic Chemistry, 34(11), 3571-3574 (1969)
T Hamaguchi et al.
British journal of cancer, 92(7), 1240-1246 (2005-03-24)
Paclitaxel (PTX) is one of the most effective anticancer agents. In clinical practice, however, high incidences of adverse reactions of the drug, for example, neurotoxicity, myelosuppression, and allergic reactions, have been reported. NK105, a micellar nanoparticle formulation, was developed to
Cyclic ether formation in oxidations of primary alcohols by cerium (IV). Reactions of 5-phenyl-1-pentanol, 4-phenyl-1-butanol, and 3-phenyl-1-propanol with ceric ammonium nitrate.
Doyle MP, et al.
The Journal of Organic Chemistry, 40(10), 1454-1456 (1975)
Min Kyung Song et al.
Journal of agricultural and food chemistry, 67(7), 2028-2035 (2019-01-31)
Caffeic acid phenethyl ester (CAPE) is an ester of a hydroxycinnamic acid (phenylpropanoid) and a phenylethanoid (2-phenylethanol; 2-PE), which has long been used in traditional medicine. Here, we synthesized 54 hydroxycinnamic acid-phenylethanoid esters by feeding 64 combinations of hydroxycinnamic acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service