Skip to Content
Merck
All Photos(3)

Key Documents

W269514

Sigma-Aldrich

(±)-2-Methylbutyric acid

greener alternative

natural, ≥98%, FG

Synonym(s):

Ethyl methyl acetic acid, Methylethyl acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)COOH
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2695
Beilstein:
1720486
EC Number:
Council of Europe no.:
2002c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.046
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.405 (lit.)

bp

176-177 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

cheesy; fruity; acidic

SMILES string

CCC(C)C(O)=O

InChI

1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

WLAMNBDJUVNPJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-2-Methylbutyric acid is one of the volatile flavor compounds identified in kogyoku apple juice and Danish Blue cheese.
Natural occurrence: Apple, apricot, cider, cocoa, cranberry, grape, grape brandy, oriental tobacco, strawberry, vinegar, whiskey.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Detection of Maize Mold Based on a Nanocomposite Colorimetric Sensor Array under Different Substrates.: This study focuses on the application of (±)-2-Methylbutyric acid in the development of nanocomposite colorimetric sensor arrays, facilitating the rapid detection of mold in maize. This advancement highlights the compound′s potential in agricultural quality control and food safety (Lin et al., 2024).

  • Regulation mechanism and bioactivity characteristic of surfactin homologues with C14 and C15 fatty acid chains.: Research elaborates on the bioactive characteristics of surfactin homologues involving (±)-2-Methylbutyric acid, underscoring its crucial role in enhancing the microbial production of bioactive compounds. This has significant implications for pharmaceuticals and bioengineering (Su et al., 2024).

  • Multifaceted analysis of the effects of roasting conditions on the flavor of fragrant Camellia oleifera Abel. seed oil.: This paper explores how (±)-2-Methylbutyric acid influences the flavor profile of Camellia oleifera seed oil under various roasting conditions, emphasizing its importance in enhancing food flavors and consumer products (Han et al., 2024).

Biochem/physiol Actions

Taste at 10 ppm

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavor components of Kogyoku apples.
Yajima I, et al.
Agricultural and Biological Chemistry, 48(4), 849-855 (1984)
Contributions of Cow, Sheep, and Goat Milks to Characterizing Branched-Chain Fatty Acid and Phenolic Flavors in Varietal Cheeses1.
Ha JK & Lindsay RC.
Journal of Dairy Science, 74(10), 3267-3274 (1991)
Kei M Igarashi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(23), 7970-7985 (2012-06-08)
Odor signals are conveyed from the olfactory bulb to the olfactory cortex (OC) by mitral cells (MCs) and tufted cells (TCs). However, whether and how the two types of projection neuron differ in function and axonal connectivity is still poorly
G Hayashida-Soiza et al.
Journal of applied microbiology, 105(5), 1672-1677 (2008-10-03)
To purify and characterize compounds with antimicrobial activity from Pseudoalteromonas haloplanktis inhibition (INH) strain. The P. haloplanktis isolated from a scallop hatchery was used to analyse antibacterial activities. Crude extracts were obtained with ethyl acetate of the cultured broth, after
Vineet K Singh et al.
Applied and environmental microbiology, 74(19), 5882-5890 (2008-08-12)
Staphylococcus aureus is a major community and nosocomial pathogen. Its ability to withstand multiple stress conditions and quickly develop resistance to antibiotics complicates the control of staphylococcal infections. Adaptation to lower temperatures is a key for the survival of bacterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service