Merck
  • Parallel mitral and tufted cell pathways route distinct odor information to different targets in the olfactory cortex.
Parallel mitral and tufted cell pathways route distinct odor information to different targets in the olfactory cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2012-06-08)
Kei M Igarashi, Nao Ieki, Myungho An, Yukie Yamaguchi, Shin Nagayama, Ko Kobayakawa, Reiko Kobayakawa, Manabu Tanifuji, Hitoshi Sakano, Wei R Chen, Kensaku Mori
ABSTRACT
Odor signals are conveyed from the olfactory bulb to the olfactory cortex (OC) by mitral cells (MCs) and tufted cells (TCs). However, whether and how the two types of projection neuron differ in function and axonal connectivity is still poorly understood. Odor responses and axonal projection patterns were compared between MCs and TCs in mice by visualizing axons of electrophysiologically identified single neurons. TCs demonstrated shorter onset latency for reliable responses than MCs. The shorter latency response of TCs was maintained in a wide range of odor concentrations, whereas MCs responded only to strong signals. Furthermore, individual TCs projected densely to focal targets only in anterior areas of the OC, whereas individual MCs dispersedly projected to all OC areas. Surprisingly, in anterior OC areas, the two cell types projected to segregated subareas. These results suggest that MCs and TCs transmit temporally distinct odor information to different OC targets.
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG