Skip to Content
Merck
All Photos(1)

Key Documents

W222216

Sigma-Aldrich

Isobutyric acid

natural, ≥99%, FCC, FG

Synonym(s):

2-Methylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2H
CAS Number:
Molecular Weight:
88.11
FEMA Number:
2222
Beilstein:
635770
EC Number:
Council of Europe no.:
6c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.006
NACRES:
NA.21
Organoleptic:
butter; acidic; rancid
grade:
FG
Fragrance grade
Kosher
natural
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

grade

FG
Fragrance grade
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

3.04 (vs air)

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

824 °F

expl. lim.

10 %

refractive index

n20/D 1.393 (lit.)

bp

153-154 °C (lit.)

mp

−47 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

butter; acidic; rancid

SMILES string

CC(C)C(O)=O

InChI

1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)

InChI key

KQNPFQTWMSNSAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Natural occurrence: Roman chamomile, strawberry, dairy products.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ritu Lal et al.
The Journal of pharmacology and experimental therapeutics, 330(3), 911-921 (2009-06-09)
Baclofen is a racemic GABA(B) receptor agonist that has a number of significant pharmacokinetic limitations, including a narrow window of absorption in the upper small intestine and rapid clearance from the blood. Arbaclofen placarbil is a novel transported prodrug of
E M Faed et al.
Clinical and experimental pharmacology & physiology, 5(2), 195-198 (1978-03-01)
1. Two main conjugates of CPIB (2-[chlorophenoxy]-2-methylpropionic acid) are present in the urine of subjects taking clofibrate. The metabolites can be separated by thin-layer chromatography (TLC). 2. Both conjugates are hydrolysed by dilute alkali, but only one is hydrolysed by
A Vyalikh et al.
Physical chemistry chemical physics : PCCP, 9(18), 2249-2257 (2007-05-10)
Solid state deuterium NMR has been used to study the molecular motion of d(6)-isobutyric acid (d(6)-iBA) in the pure (unconfined) state and confined in the cylindrical pores of two periodic mesoporous silica materials (MCM-41, pore size 3.3 nm and SBA-15
Kimberlee S Mix et al.
Molecular pharmacology, 65(2), 309-318 (2004-01-27)
Matrix metalloproteinases (MMPs) degrade extracellular matrix components, and overexpression of these enzymes contributes to tissue destruction in arthritis. Of particular importance are the collagenases, MMP-1 and MMP-13, which have high activity against the interstitial collagens in cartilage. In this study
Nicoletta Beffagna et al.
Journal of experimental botany, 58(15-16), 4183-4194 (2007-11-28)
In Arabidopsis thaliana cells, fusicoccin (FC) treatment induced an early and marked increase in the extracellular H(2)O(2) level. It also increased the huge hypo-osmotic stress-induced oxidative wave and, in addition, prevented the H(2)O(2) peak drop. These effects were apparently not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service